missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Methyldopamine hydrochloride, 95%, Thermo Scientific Chemicals
Dopamine agonist
$497.70 - $2069.08
Chemical Identifiers
CAS | 62-32-8 |
---|---|
Molecular Formula | C9H14ClNO2 |
Molecular Weight (g/mol) | 203.67 |
MDL Number | MFCD00035074 |
InChI Key | JCDRZCWRRLKLTB-UHFFFAOYSA-N |
Synonym | n-methyldopamine hydrochloride, methyldopamine hydrochloride, epinine hydrochloride, ephinine hydrochloride, deoxyepinephrine hydrochloride, unii-zv3mg8pax3, zv3mg8pax3, 3,4-dihydroxyphenylethylmethylamine hydrochloride, 4-2-methylaminoethyl pyrocatechol hydrochloride, 3,4-dihydroxyphenyl-1-methylamino-2-ethane hydrochloride |
PubChem CID | 86470 |
IUPAC Name | hydrogen 4-[2-(methylamino)ethyl]benzene-1,2-diol chloride |
SMILES | [H+].[Cl-].CNCCC1=CC=C(O)C(O)=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6030603
|
Thermo Scientific Chemicals
J6030603 |
1 g |
Each for $497.70
|
|
|||||
AAJ6030606
|
Thermo Scientific Chemicals
J6030606 |
5 g |
Each for $2,069.08
|
|
|||||
Description
Andregenic and dopamine receptor agonist; active metabolite of Ibopramine. Also, it is used as internal standard in chromatographic analysis of catecholamine's.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAndregenic and dopamine receptor agonist; active metabolite of Ibopramine. Also, it is used as internal standard in chromatographic analysis of catecholamine′s.
Solubility
Soluble in methanol, water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
Chemical Identifiers
62-32-8 | |
203.67 | |
JCDRZCWRRLKLTB-UHFFFAOYSA-N | |
86470 | |
[H+].[Cl-].CNCCC1=CC=C(O)C(O)=C1 |
C9H14ClNO2 | |
MFCD00035074 | |
n-methyldopamine hydrochloride, methyldopamine hydrochloride, epinine hydrochloride, ephinine hydrochloride, deoxyepinephrine hydrochloride, unii-zv3mg8pax3, zv3mg8pax3, 3,4-dihydroxyphenylethylmethylamine hydrochloride, 4-2-methylaminoethyl pyrocatechol hydrochloride, 3,4-dihydroxyphenyl-1-methylamino-2-ethane hydrochloride | |
hydrogen 4-[2-(methylamino)ethyl]benzene-1,2-diol chloride |
Specifications
62-32-8 | |
C9H14ClNO2 | |
1 g | |
Soluble in methanol,water. | |
[H+].[Cl-].CNCCC1=CC=C(O)C(O)=C1 | |
203.67 | |
203.67 | |
Powder |
White | |
MFCD00035074 | |
n-methyldopamine hydrochloride, methyldopamine hydrochloride, epinine hydrochloride, ephinine hydrochloride, deoxyepinephrine hydrochloride, unii-zv3mg8pax3, zv3mg8pax3, 3,4-dihydroxyphenylethylmethylamine hydrochloride, 4-2-methylaminoethyl pyrocatechol hydrochloride, 3,4-dihydroxyphenyl-1-methylamino-2-ethane hydrochloride | |
JCDRZCWRRLKLTB-UHFFFAOYSA-N | |
hydrogen 4-[2-(methylamino)ethyl]benzene-1,2-diol chloride | |
86470 | |
95% | |
N-Methyldopamine hydrochloride |
Safety and Handling
EINECSNumber : 200-528-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only