missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methylformamide 99.0+%, TCI America™

Supplier:  TCI America F0059500G

 View more versions of this product

Catalog No. F0059500G


Explore available promotions

Description

Description

Specifications

Specifications

N-Methylformamide
-5°C
183°C
MFCD00003280
methylformamide, monomethylformamide, formamide, n-methyl, n-formylmethylamine, n-methyl-formamide, n-monomethylformamide, n-methyl formamide, hconhch3, unii-xpe4g7y986, formamide, methyl
CNC=O
59.068
CHEBI:7438
≥99.0% (GC)
123-39-7
Yellow
C2H5NO
500 g
ATHHXGZTWNVVOU-UHFFFAOYSA-N
N-methylformamide
31254
59.07
Liquid

Chemical Identifiers

123-39-7
59.068
ATHHXGZTWNVVOU-UHFFFAOYSA-N
31254
N-methylformamide
C2H5NO
MFCD00003280
methylformamide, monomethylformamide, formamide, n-methyl, n-formylmethylamine, n-methyl-formamide, n-monomethylformamide, n-methyl formamide, hconhch3, unii-xpe4g7y986, formamide, methyl
CHEBI:7438
CNC=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0679

RTECSNumber : LQ3000000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions