missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Methylformanilide, 99%, Thermo Scientific Chemicals
$247.64 - $247.64
Chemical Identifiers
CAS | 93-61-8 |
---|---|
MDL Number | MFCD00003283 |
InChI Key | JIKUXBYRTXDNIY-UHFFFAOYSA-N |
Synonym | n-methylformanilide, formamide, n-methyl-n-phenyl, n-formyl-n-methylaniline, formanilide, n-methyl, methylphenylformamide, n-phenyl-n-methylformamide, n-methyl-n-formylaniline, n-methylformanilid, methyl phenyl formamide, n-methyl-n-phenyl-formamide |
PubChem CID | 66737 |
IUPAC Name | N-methyl-N-phenylformamide |
SMILES | CN(C=O)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC126810010
|
Thermo Scientific Chemicals
126810010 |
1 kg | Glass bottle |
Each for $247.64
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
93-61-8 | |
JIKUXBYRTXDNIY-UHFFFAOYSA-N | |
66737 | |
CN(C=O)C1=CC=CC=C1 |
MFCD00003283 | |
n-methylformanilide, formamide, n-methyl-n-phenyl, n-formyl-n-methylaniline, formanilide, n-methyl, methylphenylformamide, n-phenyl-n-methylformamide, n-methyl-n-formylaniline, n-methylformanilid, methyl phenyl formamide, n-methyl-n-phenyl-formamide | |
N-methyl-N-phenylformamide |
Specifications
93-61-8 | |
100.0 | |
Colorless to Yellow | |
243.0°C to 244.0°C | |
Authentic | |
Glass bottle | |
1.5600 to 1.5620 | |
MFCD00003283 | |
12,234 | |
1.09 | |
Solubility in water: immiscible. | |
CN(C=O)C1=CC=CC=C1 | |
135.17 | |
135.17 | |
Liquid |
98.5 | |
8.0°C to 13.0°C | |
1.0900g/mL | |
126°C | |
98.5% min. (GC) | |
C8H9NO | |
HCON(CH3)C6H5 | |
1 kg | |
01,68 | |
n-methylformanilide, formamide, n-methyl-n-phenyl, n-formyl-n-methylaniline, formanilide, n-methyl, methylphenylformamide, n-phenyl-n-methylformamide, n-methyl-n-formylaniline, n-methylformanilid, methyl phenyl formamide, n-methyl-n-phenyl-formamide | |
JIKUXBYRTXDNIY-UHFFFAOYSA-N | |
N-methyl-N-phenylformamide | |
66737 | |
99% | |
N-Methylformanilide, 99% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
GHS Signal Word: Warning
EINECSNumber : 202-262-3
TSCA : TSCA
RUO – Research Use Only