missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals

$131.63 - $572.40

Chemical Identifiers

CAS 4229-44-1
Molecular Formula CH6ClNO
Molecular Weight (g/mol) 83.52
MDL Number MFCD00012597
InChI Key RGZRSLKIOCHTSI-UHFFFAOYSA-N
Synonym n-methylhydroxylamine hydrochloride, methylhydroxylammonium chloride, methylhydroxylamine hydrochloride, n-methylhydroxylamine hcl, unii-mgw04sj1lz, methanamine, n-hydroxy-, hydrochloride, n-methylhydroxylammonium chloride, mgw04sj1lz, methanamine, n-hydroxy-, hydrochloride 1:1, menhoh.hcl
PubChem CID 77906
IUPAC Name N-methylhydroxylamine;hydrochloride
SMILES [H+].[Cl-].CNO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0220206
View Documents
Thermo Scientific Chemicals
L0220206
5 g
Each for $131.63
 
AAL0220214
View Documents
Thermo Scientific Chemicals
L0220214
25 g
Each for $572.40
 
Description

Description

N-Methylhydroxylamine hydrochloride is used as an inorganic catalyst used in the transamidation of primary amides with amines. It is employed in constructing hydroxamates, important functional groups for the complexation of iron.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylhydroxylamine hydrochloride is used as an inorganic catalyst used in the transamidation of primary amides with amines. It is employed in constructing hydroxamates, important functional groups for the complexation of iron.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under dry inert gas. Protect from humidity and air.
Specifications

Chemical Identifiers

4229-44-1
83.52
RGZRSLKIOCHTSI-UHFFFAOYSA-N
77906
[H+].[Cl-].CNO
CH6ClNO
MFCD00012597
n-methylhydroxylamine hydrochloride, methylhydroxylammonium chloride, methylhydroxylamine hydrochloride, n-methylhydroxylamine hcl, unii-mgw04sj1lz, methanamine, n-hydroxy-, hydrochloride, n-methylhydroxylammonium chloride, mgw04sj1lz, methanamine, n-hydroxy-, hydrochloride 1:1, menhoh.hcl
N-methylhydroxylamine;hydrochloride

Specifications

4229-44-1
CH6ClNO
5 g
Hygroscopic
Soluble in water.
[H+].[Cl-].CNO
83.52
83.52
N-Methylhydroxylamine hydrochloride
80°C to 86°C
MFCD00012597
3541409
n-methylhydroxylamine hydrochloride, methylhydroxylammonium chloride, methylhydroxylamine hydrochloride, n-methylhydroxylamine hcl, unii-mgw04sj1lz, methanamine, n-hydroxy-, hydrochloride, n-methylhydroxylammonium chloride, mgw04sj1lz, methanamine, n-hydroxy-, hydrochloride 1:1, menhoh.hcl
RGZRSLKIOCHTSI-UHFFFAOYSA-N
N-methylhydroxylamine;hydrochloride
77906
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 224-181-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only