missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methylmethacrylamide (stabilized with HQ) 98.0+%, TCI America™
Supplier: TCI America M080025ML
Specifications
| N-Methylmethacrylamide (stabilized with HQ) | |
| Yellow | |
| C5H9NO | |
| 25 mL | |
| CC(=C)C(=O)NC | |
| 99.13 | |
| 99.13 | |
| Liquid |
| 3887-02-3 | |
| 110°C | |
| MFCD00048115 | |
| WFKDPJRCBCBQNT-UHFFFAOYSA-N | |
| N,2-dimethylprop-2-enamide | |
| 77495 | |
| ≥98.0% (T) |
Chemical Identifiers
| 3887-02-3 | |
| 99.13 | |
| WFKDPJRCBCBQNT-UHFFFAOYSA-N | |
| N,2-dimethylprop-2-enamide |
| C5H9NO | |
| MFCD00048115 | |
| 77495 | |
| CC(=C)C(=O)NC |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator