Learn More
N-Methylmorpholine, 99%
CAS: 109-02-4 | C5H11NO | 101.15 g/mol
$79.42 - $432.80
Chemical Identifiers
| CAS | 109-02-4 |
|---|---|
| Molecular Formula | C5H11NO |
| Molecular Weight (g/mol) | 101.15 |
| MDL Number | MFCD00006175 |
| InChI Key | SJRJJKPEHAURKC-UHFFFAOYSA-N |
| Synonym | n-methylmorpholine, morpholine, 4-methyl, methylmorpholine, 1-methylmorpholine, morpholine, n-methyl, 4-methylmorfolin, 4-methylmorpholin, n-methyl morpholine, 4-methylmorfolin czech, n-methylmorpholin |
| PubChem CID | 7972 |
| IUPAC Name | 4-methylmorpholine |
| SMILES | CN1CCOCC1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC127151000
|
Thermo Scientific Chemicals
127151000 |
100 g | Glass Bottle |
Each for $79.42
|
|
||||
|
AC127155000
|
Thermo Scientific Chemicals
127155000 |
500 g | Glass Bottle |
Each for $113.22
|
|
||||
|
AC127150025
|
Thermo Scientific Chemicals
127150025 |
2.5 kg | Plastic drum |
Each for $432.80
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 109-02-4 | |
| 101.15 | |
| SJRJJKPEHAURKC-UHFFFAOYSA-N | |
| 7972 | |
| CN1CCOCC1 |
| C5H11NO | |
| MFCD00006175 | |
| n-methylmorpholine, morpholine, 4-methyl, methylmorpholine, 1-methylmorpholine, morpholine, n-methyl, 4-methylmorfolin, 4-methylmorpholin, n-methyl morpholine, 4-methylmorfolin czech, n-methylmorpholin | |
| 4-methylmorpholine |
Specifications
| 109-02-4 | |
| 100.0 | |
| 0.9200g/mL | |
| 14°C | |
| 98.5% min. (GC) | |
| C5H11NO | |
| MFCD00006175 | |
| 27,6 | |
| 0.92 | |
| n-methylmorpholine, morpholine, 4-methyl, methylmorpholine, 1-methylmorpholine, morpholine, n-methyl, 4-methylmorfolin, 4-methylmorpholin, n-methyl morpholine, 4-methylmorfolin czech, n-methylmorpholin | |
| SJRJJKPEHAURKC-UHFFFAOYSA-N | |
| 4-methylmorpholine | |
| 7972 | |
| 101.15 | |
| Liquid |
| 98.5 | |
| -65.0°C | |
| 115.0°C to 116.0°C (750.0 mmHg) | |
| Authentic | |
| Glass Bottle | |
| 1.4339 to 1.4359 | |
| 100 g | |
| 01,690; 02,278 | |
| 15,6362 | |
| Solubility in water: >500g/L (20°C). Other solubilities: miscible with alcohol,ether and benzene | |
| CN1CCOCC1 | |
| 101.15 | |
| 0.91 mPa.s (20°C) | |
| 99% | |
| N-Methylmorpholine, 99% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Highly flammable liquid and vapor.
GHS P Statement
Ground/Bond container and receiving equipment.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Danger
EINECSNumber : 203-640-0
RTECSNumber : QE5775000
TSCA : TSCA
RUO – Research Use Only