missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpyrrole-2-carboxaldehyde, 98%

CAS: 1192-58-1 | C6H7NO | 109.13 g/mol

$79.74 - $79.74

Chemical Identifiers

CAS 1192-58-1
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00003087
InChI Key OUKQTRFCDKSEPL-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrrole-2-carbaldehyde, 1-methylpyrrole-2-carboxaldehyde, n-methylpyrrole-2-carboxaldehyde, n-methyl-2-pyrrolecarboxaldehyde, 1-methyl-1h-pyrrole-2-carboxaldehyde, 2-formyl-1-methylpyrrole, 1-methyl-2-pyrrolecarboxaldehyde, 1h-pyrrole-2-carboxaldehyde, 1-methyl, unii-m0hyh3d7sx, 1-methyl-2-formylpyrrole
PubChem CID 14504
IUPAC Name 1-methylpyrrole-2-carbaldehyde
SMILES CN1C=CC=C1C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC156930050
SDS View Documents
Thermo Scientific Chemicals
156930050
5 g Glass bottle
Each for $79.74
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1192-58-1
109.13
OUKQTRFCDKSEPL-UHFFFAOYSA-N
14504
CN1C=CC=C1C=O
C6H7NO
MFCD00003087
1-methyl-1h-pyrrole-2-carbaldehyde, 1-methylpyrrole-2-carboxaldehyde, n-methylpyrrole-2-carboxaldehyde, n-methyl-2-pyrrolecarboxaldehyde, 1-methyl-1h-pyrrole-2-carboxaldehyde, 2-formyl-1-methylpyrrole, 1-methyl-2-pyrrolecarboxaldehyde, 1h-pyrrole-2-carboxaldehyde, 1-methyl, unii-m0hyh3d7sx, 1-methyl-2-formylpyrrole
1-methylpyrrole-2-carbaldehyde

Specifications

1192-58-1
100.0
87°C to 90°C (22.0 mmHg)
Authentic
Glass bottle
1.5580 to 1.562
5 g
1.01
OUKQTRFCDKSEPL-UHFFFAOYSA-N
1-methylpyrrole-2-carbaldehyde
14504
98%
N-Methylpyrrole-2-carboxaldehyde
97.5
1.0100g/mL
72°C
97.5% min. (GC)
C6H7NO
MFCD00003087
21, I, 279
1-methyl-1h-pyrrole-2-carbaldehyde, 1-methylpyrrole-2-carboxaldehyde, n-methylpyrrole-2-carboxaldehyde, n-methyl-2-pyrrolecarboxaldehyde, 1-methyl-1h-pyrrole-2-carboxaldehyde, 2-formyl-1-methylpyrrole, 1-methyl-2-pyrrolecarboxaldehyde, 1h-pyrrole-2-carboxaldehyde, 1-methyl, unii-m0hyh3d7sx, 1-methyl-2-formylpyrrole
CN1C=CC=C1C=O
109.13
109.13
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 214-755-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only