missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Bis(2-hydroxyethyl)ethylenediamine, Thermo Scientific Chemicals

$184.96 - $732.37

Chemical Identifiers

CAS 3197-06-6
Molecular Formula C6H16N2O2
Molecular Weight (g/mol) 148.21
MDL Number MFCD00014822
InChI Key CYOIAXUAIXVWMU-UHFFFAOYSA-N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3231203
View Documents
Thermo Scientific Chemicals
03231203
1 g
Each for $184.96
 
AA3231206
View Documents
Thermo Scientific Chemicals
03231206
5 g
Each for $732.37
 
Description

Description

It is employed as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3197-06-6
148.21
CYOIAXUAIXVWMU-UHFFFAOYSA-N
C6H16N2O2
MFCD00014822

Specifications

3197-06-6
174°C to 178°C (3 to 4 mmHg)
C6H16N2O2
1 g
n,n-bis 2-hydroxyethyl ethylenediamine, 2,2'-2-aminoethyl azanediyl diethanol, ethanol, 2,2'-2-aminoethyl imino bis, 2,2'-2-aminoethyl imino bisethanol, 2-2-aminoethyl 2-hydroxyethyl amino ethanol, 2-2-aminoethyl 2-hydroxyethyl amino ethan-1-ol, aminoethyldiethanolamine, aminoethylethanolamine solution, 2,2'-2-aminoethyl imino diethanol, acmc-20a9el
CYOIAXUAIXVWMU-UHFFFAOYSA-N
2-[2-aminoethyl(2-hydroxyethyl)amino]ethanol
98535
Liquid
1.069 g/mL
Amine-like
MFCD00014822
UN2735
Soluble in water.
C(CN(CCO)CCO)N
148.21
148.21
N,N-Bis(2-hydroxyethyl)ethylenediamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only