Learn More
N-(n-Butyl)aniline, 99%, Thermo Scientific Chemicals
$129.95 - $383.84
Chemical Identifiers
CAS | 1126-78-9 |
---|---|
Molecular Formula | C10H15N |
Molecular Weight (g/mol) | 149.237 |
MDL Number | MFCD00035798 |
InChI Key | VSHTWPWTCXQLQN-UHFFFAOYSA-N |
Synonym | benzenamine, n-butyl, aniline, n-butyl, 4-phenylamino butane, n-butylbenzenamine, n-n-butyl aniline, butylaniline, n-phenyl-n-butylamine, butylphenylamine, unii-r2znj7l2um, benzamine, n-butyl |
PubChem CID | 14310 |
IUPAC Name | N-butylaniline |
SMILES | CCCCNC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2418014
|
Thermo Scientific Chemicals
B2418014 |
25 g |
Each for $129.95
|
|
|||||
AAB2418022
|
Thermo Scientific Chemicals
B2418022 |
100 g |
Each for $383.84
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1126-78-9 | |
149.237 | |
VSHTWPWTCXQLQN-UHFFFAOYSA-N | |
14310 | |
CCCCNC1=CC=CC=C1 |
C10H15N | |
MFCD00035798 | |
benzenamine, n-butyl, aniline, n-butyl, 4-phenylamino butane, n-butylbenzenamine, n-n-butyl aniline, butylaniline, n-phenyl-n-butylamine, butylphenylamine, unii-r2znj7l2um, benzamine, n-butyl | |
N-butylaniline |
Specifications
1126-78-9 | |
240°C to 242°C | |
C10H15N | |
MFCD00035798 | |
UN2738 | |
VSHTWPWTCXQLQN-UHFFFAOYSA-N | |
N-butylaniline | |
14310 | |
99% |
0.932 | |
105°C (222°F) | |
1.532 | |
25 g | |
benzenamine, n-butyl, aniline, n-butyl, 4-phenylamino butane, n-butylbenzenamine, n-n-butyl aniline, butylaniline, n-phenyl-n-butylamine, butylphenylamine, unii-r2znj7l2um, benzamine, n-butyl | |
CCCCNC1=CC=CC=C1 | |
149.237 | |
149.23 | |
N-(n-Butyl)aniline |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: N-BUTYLANILINE
EINECSNumber : 214-425-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only