missing translation for 'onlineSavingsMsg'
Learn More

N-(n-Butyl)ethylenediamine, 97%

CAS: 19522-69-1 | C6H16N2 | 116.208 g/mol

$181.82 - $730.84

Chemical Identifiers

CAS 19522-69-1
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.208
MDL Number MFCD00025602
InChI Key DFPGBRPWDZFIPP-UHFFFAOYSA-N
Synonym n-butylethylenediamine, 2-butylamino ethylamine, n1-butylethane-1,2-diamine, 2-aminoethyl butyl amine, 1,2-ethanediamine,n1-butyl, 2-butylamino-ethylamine, 2-n-butylaminoethylamine, acmc-1bs5m, n-butyl-1,2-ethanediamine, n-butylethane-1,2-diamine
PubChem CID 519668
IUPAC Name N'-butylethane-1,2-diamine
SMILES CCCCNCCN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1343706
View Documents
Thermo Scientific Chemicals
L1343706
5 g N/A
Only null left
 
AAL1343714
View Documents
Thermo Scientific Chemicals
L1343714
25 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

19522-69-1
116.208
DFPGBRPWDZFIPP-UHFFFAOYSA-N
519668
CCCCNCCN
C6H16N2
MFCD00025602
n-butylethylenediamine, 2-butylamino ethylamine, n1-butylethane-1,2-diamine, 2-aminoethyl butyl amine, 1,2-ethanediamine,n1-butyl, 2-butylamino-ethylamine, 2-n-butylaminoethylamine, acmc-1bs5m, n-butyl-1,2-ethanediamine, n-butylethane-1,2-diamine
N'-butylethane-1,2-diamine

Specifications

19522-69-1
170°C to 171°C
C6H16N2
MFCD00025602
UN2734
Air Sensitive
DFPGBRPWDZFIPP-UHFFFAOYSA-N
N'-butylethane-1,2-diamine
519668
97%
0.836
59°C (138°F)
1.443
5 g
1732700
n-butylethylenediamine, 2-butylamino ethylamine, n1-butylethane-1,2-diamine, 2-aminoethyl butyl amine, 1,2-ethanediamine,n1-butyl, 2-butylamino-ethylamine, 2-n-butylaminoethylamine, acmc-1bs5m, n-butyl-1,2-ethanediamine, n-butylethane-1,2-diamine
CCCCNCCN
116.208
116.21
N-(n-Butyl)ethylenediamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H226-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only