missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-N-Butyldiethanolamine, 98%, Thermo Scientific Chemicals

$62.75 - $328.11

Chemical Identifiers

CAS 102-79-4
Molecular Formula C8H19NO2
Molecular Weight (g/mol) 161.24
MDL Number MFCD00002856
InChI Key GVNHOISKXMSMPX-UHFFFAOYSA-N
Synonym n-butyldiethanolamine, butyldiethanolamine, ethanol, 2,2'-butylimino bis, bide, 2,2'-butylimino diethanol, n,n-bis 2-hydroxyethyl butylamine, butylbis 2-hydroxyethyl amine, n-butyl-2,2'-iminodiethanol, 2,2-butylimino diethanol, n-butyl-n,n-bis hydroxyethyl amine
PubChem CID 7620
IUPAC Name 2-[butyl(2-hydroxyethyl)amino]ethanol
SMILES CCCCN(CCO)CCO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC154362500
SDS View Documents
Thermo Scientific Chemicals
154362500
250 mL Glass bottle
Each for $62.75
 
AC154360010
SDS View Documents
Thermo Scientific Chemicals
154360010
1 L Glass bottle
Each for $153.68
 
AC154360025
SDS View Documents
Thermo Scientific Chemicals
154360025
2.5 L Glass bottle
Each for $328.11
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

102-79-4
161.24
GVNHOISKXMSMPX-UHFFFAOYSA-N
7620
CCCCN(CCO)CCO
C8H19NO2
MFCD00002856
n-butyldiethanolamine, butyldiethanolamine, ethanol, 2,2'-butylimino bis, bide, 2,2'-butylimino diethanol, n,n-bis 2-hydroxyethyl butylamine, butylbis 2-hydroxyethyl amine, n-butyl-2,2'-iminodiethanol, 2,2-butylimino diethanol, n-butyl-n,n-bis hydroxyethyl amine
2-[butyl(2-hydroxyethyl)amino]ethanol

Specifications

102-79-4
100.0
Brown to Brown-Red
264.0°C to 278.0°C
Authentic
Glass bottle
1.4615 to 1.4635
MFCD00002856
04, 285
n-butyldiethanolamine, butyldiethanolamine, ethanol, 2,2'-butylimino bis, bide, 2,2'-butylimino diethanol, n,n-bis 2-hydroxyethyl butylamine, butylbis 2-hydroxyethyl amine, n-butyl-2,2'-iminodiethanol, 2,2-butylimino diethanol, n-butyl-n,n-bis hydroxyethyl amine
GVNHOISKXMSMPX-UHFFFAOYSA-N
2-[butyl(2-hydroxyethyl)amino]ethanol
7620
98%
N-N-Butyldiethanolamine
97.5
-70.0°C
0.9680g/mL
143°C
97.5% min. (GC)
C8H19NO2
CH3(CH2)3N(CH2CH2OH)2
250 mL
0.968
Solubility in water: miscible
CCCCN(CCO)CCO
161.24
161.24
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-055-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only