missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-n-Butyldiethanolamine, 98%, Thermo Scientific Chemicals

$51.88 - $130.19

Chemical Identifiers

CAS 102-79-4
Molecular Formula C8H19NO2
Molecular Weight (g/mol) 161.245
MDL Number MFCD00002856
InChI Key GVNHOISKXMSMPX-UHFFFAOYSA-N
Synonym n-butyldiethanolamine, butyldiethanolamine, ethanol, 2,2'-butylimino bis, bide, 2,2'-butylimino diethanol, n,n-bis 2-hydroxyethyl butylamine, butylbis 2-hydroxyethyl amine, n-butyl-2,2'-iminodiethanol, 2,2-butylimino diethanol, n-butyl-n,n-bis hydroxyethyl amine
PubChem CID 7620
IUPAC Name 2-[butyl(2-hydroxyethyl)amino]ethanol
SMILES CCCCN(CCO)CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0995322
View Documents
Thermo Scientific Chemicals
L0995322
100 g
Each for $51.88
 
AAL0995336
View Documents
Thermo Scientific Chemicals
L0995336
500 g
Each for $130.19
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

102-79-4
161.245
GVNHOISKXMSMPX-UHFFFAOYSA-N
7620
CCCCN(CCO)CCO
C8H19NO2
MFCD00002856
n-butyldiethanolamine, butyldiethanolamine, ethanol, 2,2'-butylimino bis, bide, 2,2'-butylimino diethanol, n,n-bis 2-hydroxyethyl butylamine, butylbis 2-hydroxyethyl amine, n-butyl-2,2'-iminodiethanol, 2,2-butylimino diethanol, n-butyl-n,n-bis hydroxyethyl amine
2-[butyl(2-hydroxyethyl)amino]ethanol

Specifications

102-79-4
0.968
143°C (289°F)
1.464
100 g
1739642
n-butyldiethanolamine, butyldiethanolamine, ethanol, 2,2'-butylimino bis, bide, 2,2'-butylimino diethanol, n,n-bis 2-hydroxyethyl butylamine, butylbis 2-hydroxyethyl amine, n-butyl-2,2'-iminodiethanol, 2,2-butylimino diethanol, n-butyl-n,n-bis hydroxyethyl amine
CCCCN(CCO)CCO
161.245
161.25
N-n-Butyldiethanolamine
-70°C
268°C to 272°C
C8H19NO2
MFCD00002856
UN2735
Hygroscopic
GVNHOISKXMSMPX-UHFFFAOYSA-N
2-[butyl(2-hydroxyethyl)amino]ethanol
7620
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 203-055-0

RTECSNumber : KK0525000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only