missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N,N'-Dibenzoylhydrazine 95.0+%, TCI America™
$128.38 - $396.97
Chemical Identifiers
CAS | 787-84-8 |
---|---|
Molecular Formula | C14H12N2O2 |
Molecular Weight (g/mol) | 240.262 |
MDL Number | MFCD00003068 |
InChI Key | GRRIYLZJLGTQJX-UHFFFAOYSA-N |
Synonym | dibenzoylhydrazine, n,n'-dibenzoylhydrazine, 1,2-dibenzoylhydrazine, benzoic acid, 2-benzoylhydrazide, n2-benzoylbenzoic acid hydrazide, hydrazine, 1,2-dibenzoyl, n,n-dibenzoylhydrazine, nn'-dibenzoylhydrazine, c6h5-co-nh-nh-co-c6h5, benzoic acid,2-benzoylhydrazide |
PubChem CID | 13085 |
ChEBI | CHEBI:38453 |
IUPAC Name | N'-benzoylbenzohydrazide |
SMILES | C1=CC=C(C=C1)C(=O)NNC(=O)C2=CC=CC=C2 |
Chemical Identifiers
787-84-8 | |
240.262 | |
GRRIYLZJLGTQJX-UHFFFAOYSA-N | |
13085 | |
N'-benzoylbenzohydrazide |
C14H12N2O2 | |
MFCD00003068 | |
dibenzoylhydrazine, n,n'-dibenzoylhydrazine, 1,2-dibenzoylhydrazine, benzoic acid, 2-benzoylhydrazide, n2-benzoylbenzoic acid hydrazide, hydrazine, 1,2-dibenzoyl, n,n-dibenzoylhydrazine, nn'-dibenzoylhydrazine, c6h5-co-nh-nh-co-c6h5, benzoic acid,2-benzoylhydrazide | |
CHEBI:38453 | |
C1=CC=C(C=C1)C(=O)NNC(=O)C2=CC=CC=C2 |
Specifications
787-84-8 | |
C14H12N2O2 | |
5 g | |
GRRIYLZJLGTQJX-UHFFFAOYSA-N | |
N'-benzoylbenzohydrazide | |
13085 | |
240.26 | |
Crystalline Powder |
White-Yellow | |
MFCD00003068 | |
dibenzoylhydrazine, n,n'-dibenzoylhydrazine, 1,2-dibenzoylhydrazine, benzoic acid, 2-benzoylhydrazide, n2-benzoylbenzoic acid hydrazide, hydrazine, 1,2-dibenzoyl, n,n-dibenzoylhydrazine, nn'-dibenzoylhydrazine, c6h5-co-nh-nh-co-c6h5, benzoic acid,2-benzoylhydrazide | |
C1=CC=C(C=C1)C(=O)NNC(=O)C2=CC=CC=C2 | |
240.262 | |
CHEBI:38453 | |
≥95.0% (T) | |
N,N′-Dibenzoylhydrazine |
Safety and Handling
RTECSNumber : MV2085000
TSCA : Yes