missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N,N'-Dicyclohexylmorpholine-4-carboxamidine, 98+%, Thermo Scientific™
$216.56 - $216.56
Chemical Identifiers
CAS | 4975-73-9 |
---|---|
Molecular Formula | C17H31N3O |
Molecular Weight (g/mol) | 293.455 |
MDL Number | MFCD00063252 |
InChI Key | OZNYZQOTXQSUJM-UHFFFAOYSA-N |
Synonym | n,n'-dicyclohexyl-4-morpholinecarboxamidine, n,n'-dicyclohexyl-4-morpholine carboxamidine, n,n'-dicyclohexyl-1-morpholinoformamidine, n,n'-dicyclohexylmorpholine-4-carboxamidine, 4-morpholinecarboximidamide, n,n'-dicyclohexyl, formamidine, n,n'-dicyclohexyl-1-morpholino, n,n'-dicyclohexyl-4-morpholine-carboxamidine, z-n,n'-dicyclohexylmorpholine-4-carboximidamide, 4-n,n'-dicyclohexylamidino morpholine, n,n'-dicyclohexyl-4-morpholinecarboximidamide |
PubChem CID | 21079 |
IUPAC Name | N,N'-dicyclohexylmorpholine-4-carboximidamide |
SMILES | C1CCC(CC1)NC(=NC2CCCCC2)N3CCOCC3 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5648014
|
Thermo Scientific Chemicals
H5648014 |
25 g |
Each for $216.56
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
4975-73-9 | |
293.455 | |
OZNYZQOTXQSUJM-UHFFFAOYSA-N | |
21079 | |
C1CCC(CC1)NC(=NC2CCCCC2)N3CCOCC3 |
C17H31N3O | |
MFCD00063252 | |
n,n'-dicyclohexyl-4-morpholinecarboxamidine, n,n'-dicyclohexyl-4-morpholine carboxamidine, n,n'-dicyclohexyl-1-morpholinoformamidine, n,n'-dicyclohexylmorpholine-4-carboxamidine, 4-morpholinecarboximidamide, n,n'-dicyclohexyl, formamidine, n,n'-dicyclohexyl-1-morpholino, n,n'-dicyclohexyl-4-morpholine-carboxamidine, z-n,n'-dicyclohexylmorpholine-4-carboximidamide, 4-n,n'-dicyclohexylamidino morpholine, n,n'-dicyclohexyl-4-morpholinecarboximidamide | |
N,N'-dicyclohexylmorpholine-4-carboximidamide |
Specifications
4975-73-9 | |
C17H31N3O | |
25 g | |
OZNYZQOTXQSUJM-UHFFFAOYSA-N | |
N,N'-dicyclohexylmorpholine-4-carboximidamide | |
21079 | |
≥98% |
105°C to 107°C | |
MFCD00063252 | |
n,n'-dicyclohexyl-4-morpholinecarboxamidine, n,n'-dicyclohexyl-4-morpholine carboxamidine, n,n'-dicyclohexyl-1-morpholinoformamidine, n,n'-dicyclohexylmorpholine-4-carboxamidine, 4-morpholinecarboximidamide, n,n'-dicyclohexyl, formamidine, n,n'-dicyclohexyl-1-morpholino, n,n'-dicyclohexyl-4-morpholine-carboxamidine, z-n,n'-dicyclohexylmorpholine-4-carboximidamide, 4-n,n'-dicyclohexylamidino morpholine, n,n'-dicyclohexyl-4-morpholinecarboximidamide | |
C1CCC(CC1)NC(=NC2CCCCC2)N3CCOCC3 | |
293.455 | |
293.45 | |
N,N'-Dicyclohexylmorpholine-4-carboxamidine |
Safety and Handling
EINECSNumber : 225-622-1
RTECSNumber : LQ4566500
TSCA : No
Recommended Storage : Ambient temperatures