Learn More
N,N-Diethyl-3-methylbenzamide, 97%, Thermo Scientific Chemicals
$61.55 - $445.59
Chemical Identifiers
CAS | 134-62-3 |
---|---|
Molecular Formula | C12H17NO |
Molecular Weight (g/mol) | 191.274 |
MDL Number | MFCD00009046 |
InChI Key | MMOXZBCLCQITDF-UHFFFAOYSA-N |
Synonym | deet, n,n-diethyl-m-toluamide, diethyltoluamide, dieltamid, metadelphene, flypel, delphene, deta, detamide, autan |
PubChem CID | 4284 |
ChEBI | CHEBI:7071 |
IUPAC Name | N,N-diethyl-3-methylbenzamide |
SMILES | CCN(CC)C(=O)C1=CC(=CC=C1)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1748622
|
Thermo Scientific Chemicals
A1748622 |
100 g |
Each for $61.55
|
|
|||||
AAA1748636
|
Thermo Scientific Chemicals
A1748636 |
500 g |
Each for $135.14
|
|
|||||
AAA174860E
|
Thermo Scientific Chemicals
A174860E |
2500 g |
Each for $445.59
|
|
|||||
Description
It is the most common ingredient in insect repellants and is believed to work as such in that mosquitoes intensely dislike its smell. Deet is also an effective solvent and may dissolve plastics, rayon, spandex, other synthetic fabrics and painted or varnished surfaces.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is the most common ingredient in insect repellants and is believed to work as such in that mosquitoes intensely dislike its smell. Deet is also an effective solvent and may dissolve plastics, rayon, spandex, other synthetic fabrics and painted or varnished surfaces.
Solubility
Soluble in water.
Notes
Keep away from strong oxidizing agents, strong acids, strong bases, strong reducing agents. Keep ate room temperature.
Chemical Identifiers
134-62-3 | |
191.274 | |
MMOXZBCLCQITDF-UHFFFAOYSA-N | |
4284 | |
N,N-diethyl-3-methylbenzamide |
C12H17NO | |
MFCD00009046 | |
deet, n,n-diethyl-m-toluamide, diethyltoluamide, dieltamid, metadelphene, flypel, delphene, deta, detamide, autan | |
CHEBI:7071 | |
CCN(CC)C(=O)C1=CC(=CC=C1)C |
Specifications
134-62-3 | |
110°C to 112°C (1 mmHg) | |
C12H17NO | |
MFCD00009046 | |
2046711 | |
deet, n,n-diethyl-m-toluamide, diethyltoluamide, dieltamid, metadelphene, flypel, delphene, deta, detamide, autan | |
MMOXZBCLCQITDF-UHFFFAOYSA-N | |
N,N-diethyl-3-methylbenzamide | |
4284 | |
191.27 | |
N,N-Diethyl-3-methylbenzamide |
0.997 | |
145°C (293°F) | |
1.523 | |
100 g | |
14,2856 | |
Soluble in water. | |
CCN(CC)C(=O)C1=CC(=CC=C1)C | |
191.274 | |
CHEBI:7071 | |
97% |
Safety and Handling
GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H302-H315-H319-H336-H373
EINECSNumber : 205-149-7
RTECSNumber : XS3675000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only