missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Diethyl-m-toluamide, 98%, Thermo Scientific Chemicals

$54.70 - $128.20

Chemical Identifiers

CAS 134-62-3
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.27
MDL Number MFCD00009046
InChI Key MMOXZBCLCQITDF-UHFFFAOYSA-N
Synonym deet, n,n-diethyl-m-toluamide, diethyltoluamide, dieltamid, metadelphene, flypel, delphene, deta, detamide, autan
PubChem CID 4284
ChEBI CHEBI:7071
IUPAC Name N,N-diethyl-3-methylbenzamide
SMILES CCN(CC)C(=O)C1=CC(=CC=C1)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC114571000
SDS View Documents
Thermo Scientific Chemicals
114571000
100 g Glass bottle
Each for $54.70
 
AC114575000
SDS View Documents
Thermo Scientific Chemicals
114575000
500 g Glass bottle
Each for $128.20
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

134-62-3
191.27
MMOXZBCLCQITDF-UHFFFAOYSA-N
4284
N,N-diethyl-3-methylbenzamide
C12H17NO
MFCD00009046
deet, n,n-diethyl-m-toluamide, diethyltoluamide, dieltamid, metadelphene, flypel, delphene, deta, detamide, autan
CHEBI:7071
CCN(CC)C(=O)C1=CC(=CC=C1)C

Specifications

134-62-3
100.0
111°C (1.0 mmHg)
Authentic
Glass bottle
1.5200 to 1.524
MFCD00009046
09, II, 325
15, 2859
Solubility in water: negligible. Other solubilities: soluble in ethanol,chloroform,diethyl ether,,benzene
CCN(CC)C(=O)C1=CC(=CC=C1)C
191.27
13.3 mPa.s (30°C)
191.27
Liquid
97.5
0.9955g/mL
155°C
97.5% min. (GC)
C12H17NO
CH3C6H4CON(C2H5)2
100 g
0.9955
deet, n,n-diethyl-m-toluamide, diethyltoluamide, dieltamid, metadelphene, flypel, delphene, deta, detamide, autan
MMOXZBCLCQITDF-UHFFFAOYSA-N
N,N-diethyl-3-methylbenzamide
4284
CHEBI:7071
98%
N, N-Diethyl-m-toluamide, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.

GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 205-149-7

RTECSNumber : XS3675000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only