missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N,N-Diethyl-m-toluamide 98.0+%, TCI America™
$65.17 - $242.92
Chemical Identifiers
CAS | 134-62-3 |
---|---|
Molecular Formula | C12H17NO |
Molecular Weight (g/mol) | 191.274 |
MDL Number | MFCD00009046 |
InChI Key | MMOXZBCLCQITDF-UHFFFAOYSA-N |
Synonym | deet, n,n-diethyl-m-toluamide, diethyltoluamide, dieltamid, metadelphene, flypel, delphene, deta, detamide, autan |
PubChem CID | 4284 |
ChEBI | CHEBI:7071 |
IUPAC Name | N,N-diethyl-3-methylbenzamide |
SMILES | CCN(CC)C(=O)C1=CC(=CC=C1)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
D009725ML
|
TCI America
D009725ML |
25 mL |
Each for $65.17
|
|
|||||
D0097500ML
|
TCI America
D0097500ML |
500 mL |
Each for $242.92
|
|
|||||
Chemical Identifiers
134-62-3 | |
191.274 | |
MMOXZBCLCQITDF-UHFFFAOYSA-N | |
4284 | |
N,N-diethyl-3-methylbenzamide |
C12H17NO | |
MFCD00009046 | |
deet, n,n-diethyl-m-toluamide, diethyltoluamide, dieltamid, metadelphene, flypel, delphene, deta, detamide, autan | |
CHEBI:7071 | |
CCN(CC)C(=O)C1=CC(=CC=C1)C |
Specifications
134-62-3 | |
160°C | |
MFCD00009046 | |
deet, n,n-diethyl-m-toluamide, diethyltoluamide, dieltamid, metadelphene, flypel, delphene, deta, detamide, autan | |
CCN(CC)C(=O)C1=CC(=CC=C1)C | |
191.274 | |
CHEBI:7071 | |
≥98.0% (GC) | |
N,N-Diethyl-m-toluamide |
Yellow | |
C12H17NO | |
25 mL | |
MMOXZBCLCQITDF-UHFFFAOYSA-N | |
N,N-diethyl-3-methylbenzamide | |
4284 | |
191.27 | |
Liquid |
Safety and Handling
EINECSNumber : (3)-1321&(3)-1677
RTECSNumber : XS3675000
TSCA : Yes