Learn More
N,N-Diethyl-p-phenylenediamine sulfate, 99%, Thermo Scientific Chemicals
N, N-Diethyl-p-phenylenediamine sulfate, 99%, C10H16N2.H2SO4, CAS Number-6283-63-2
Supplier: Thermo Scientific Chemicals 395000010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N, N-Diethyl-p-phenylenediamine sulfate | |
98.5 | |
184.0°C to 187.0°C | |
Authentic | |
Glass bottle | |
MFCD00012993 | |
n,n-diethyl-p-phenylenediamine sulfate, n1,n1-diethylbenzene-1,4-diamine sulfate, 4-amino-n,n-diethylaniline sulfate, diethyl-p-phenylenediamine sulfate, n,n-diethyl-1,4-phenylenediamine sulfate, unii-usp19t3gda, 1,4-benzenediamine, n,n-diethyl-, sulfate 1:1, 1,4-benzenediamine, n,n-diethyl-, sulfate, n,n-diethyl-1,4-benzenediamine sulfate, p-phenylenediamine, n,n-diethyl-, sulfate 1:1 | |
CCN(CC)C1=CC=C(C=C1)N.OS(=O)(=O)O | |
262.33 | |
262.33 | |
Crystalline Powder or Crystals With Lumps |
6283-63-2 | |
100.0 | |
White to Beige | |
99% | |
C10H16N2·H2SO4 | |
1 kg | |
AYLDJQABCMPYEN-UHFFFAOYSA-N | |
4-N,4-N-diethylbenzene-1,4-diamine;sulfuric acid | |
80166 | |
99% |
Chemical Identifiers
6283-63-2 | |
262.33 | |
AYLDJQABCMPYEN-UHFFFAOYSA-N | |
80166 | |
CCN(CC)C1=CC=C(C=C1)N.OS(=O)(=O)O |
C10H16N2·H2SO4 | |
MFCD00012993 | |
n,n-diethyl-p-phenylenediamine sulfate, n1,n1-diethylbenzene-1,4-diamine sulfate, 4-amino-n,n-diethylaniline sulfate, diethyl-p-phenylenediamine sulfate, n,n-diethyl-1,4-phenylenediamine sulfate, unii-usp19t3gda, 1,4-benzenediamine, n,n-diethyl-, sulfate 1:1, 1,4-benzenediamine, n,n-diethyl-, sulfate, n,n-diethyl-1,4-benzenediamine sulfate, p-phenylenediamine, n,n-diethyl-, sulfate 1:1 | |
4-N,4-N-diethylbenzene-1,4-diamine;sulfuric acid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
Wear protective gloves/protective clothing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
GHS Signal Word: Warning
EINECSNumber : 228-500-6
RUO – Research Use Only