Learn More
N,N-Diethylaniline, 99%
CAS: 91-66-7 | C10H15N | 149.24 g/mol
$57.22 - $271.88
Chemical Identifiers
| CAS | 91-66-7 |
|---|---|
| Molecular Formula | C10H15N |
| Molecular Weight (g/mol) | 149.24 |
| MDL Number | MFCD00009042,MFCD31699978 |
| InChI Key | GGSUCNLOZRCGPQ-UHFFFAOYSA-N |
| Synonym | diethylaniline, n,n-diethylanilin, diethylphenylamine, benzenamine, n,n-diethyl, n,n-diethyl aniline, diaethylanilin, phenyldiethylamine, aniline, n,n-diethyl, n,n-diethylaminobenzene, n-phenyldiethylamine |
| PubChem CID | 7061 |
| IUPAC Name | N,N-diethylaniline |
| SMILES | CCN(CC)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC181062500
|
Thermo Scientific Chemicals
181062500 |
250 mL | Glass bottle |
N/A
|
|
||||
|
AC181060010
|
Thermo Scientific Chemicals
181060010 |
1 L | Glass bottle |
N/A
|
|
||||
|
AC181060025
|
Thermo Scientific Chemicals
181060025 |
2.5 L | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 91-66-7 | |
| 149.24 | |
| GGSUCNLOZRCGPQ-UHFFFAOYSA-N | |
| 7061 | |
| CCN(CC)C1=CC=CC=C1 |
| C10H15N | |
| MFCD00009042,MFCD31699978 | |
| diethylaniline, n,n-diethylanilin, diethylphenylamine, benzenamine, n,n-diethyl, n,n-diethyl aniline, diaethylanilin, phenyldiethylamine, aniline, n,n-diethyl, n,n-diethylaminobenzene, n-phenyldiethylamine | |
| N,N-diethylaniline |
Specifications
| 91-66-7 | |
| 100.0 | |
| Yellow | |
| 215.0°C to 217.0°C | |
| Authentic | |
| Glass bottle | |
| 1.5410 to 1.5430 | |
| MFCD00009042,MFCD31699978 | |
| 12, 164 | |
| 0.933 | |
| diethylaniline, n,n-diethylanilin, diethylphenylamine, benzenamine, n,n-diethyl, n,n-diethyl aniline, diaethylanilin, phenyldiethylamine, aniline, n,n-diethyl, n,n-diethylaminobenzene, n-phenyldiethylamine | |
| GGSUCNLOZRCGPQ-UHFFFAOYSA-N | |
| N,N-diethylaniline | |
| 7061 | |
| 99% | |
| N, N-Diethylaniline, 99% |
| 98.5 | |
| -38.0°C | |
| 0.9330g/mL | |
| 84°C | |
| 99% | |
| C10H15N | |
| C6H5N(C2H5)2 | |
| 250 mL | |
| 05,212; 16,182 | |
| 15, 3125 | |
| Solubility in water: 10-27g/L. Other solubilities: slightly soluble in alcohol,chloroform and ether | |
| CCN(CC)C1=CC=CC=C1 | |
| 149.24 | |
| 149.24 | |
| Liquid |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.
Toxic if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
GHS Signal Word: Danger
EINECSNumber : 202-088-8
RTECSNumber : BX3400000
TSCA : TSCA
RUO – Research Use Only