missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N,N-Diisopropylethylamine (Peptide Synthesis), Fisher BioReagents
Supplier: Fisher BioReagents BP592500
Description
N,N-Diisopropylethylamine is commonly used in peptide synthesis reactions.Specifications
Diisopropylethylamine | |
7087-68-5 | |
Yellow | |
127°C | |
Glass Bottle | |
1.4113 to 1.4153 (at 25°C) | |
15, 3219 | |
JGFZNNIVVJXRND-UHFFFAOYSA-N | |
N-ethyl-N-propan-2-ylpropan-2-amine | |
81531 | |
38mmHg at 31°C | |
Peptide Synthesis |
N, N-Diisopropylethylamine | |
-46°C | |
0.7560g/cm³ | |
≥99 % | |
C8H19N | |
500 mL | |
n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine | |
CCN(C(C)C)C(C)C | |
129.247 | |
129.24 | |
≥99% | |
Liquid |
Chemical Identifiers
7087-68-5 | |
129.247 | |
n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine | |
N-ethyl-N-propan-2-ylpropan-2-amine |
C8H19N | |
JGFZNNIVVJXRND-UHFFFAOYSA-N | |
81531 | |
CCN(C(C)C)C(C)C |
Safety and Handling
Recommended Storage : RT