missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diisopropylethylenediamine 97.0+%, TCI America™

$89.48 - $922.71

Chemical Identifiers

CAS 121-05-1
Molecular Formula C8H20N2
Molecular Weight (g/mol) 144.262
MDL Number MFCD00015015
InChI Key CURJNMSGPBXOGK-UHFFFAOYSA-N
Synonym 2-aminoethyldiisopropylamine, n,n-diisopropylethylenediamine, 2-diisopropylamino ethylamine, usaf am-2, 1,2-ethanediamine, n,n-bis 1-methylethyl, n,n-diisopropyl-1,2-ethanediamine, n1,n1-diisopropylethane-1,2-diamine, 2-aminoethyl diisopropylamine, n,n-diisopropyl ethylenediamine, 2-diisopropylaminoethylamine
PubChem CID 8459
IUPAC Name N',N'-di(propan-2-yl)ethane-1,2-diamine
SMILES CC(C)N(CCN)C(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D000625ML
View Documents
TCI America
D000625ML
25 mL
Each for $89.48
Only null left
 
D0006500ML
View Documents
TCI America
D0006500ML
500 mL
Each for $922.71
Only null left
 
Description

Description

Specifications

Chemical Identifiers

121-05-1
144.262
CURJNMSGPBXOGK-UHFFFAOYSA-N
8459
CC(C)N(CCN)C(C)C
C8H20N2
MFCD00015015
2-aminoethyldiisopropylamine, n,n-diisopropylethylenediamine, 2-diisopropylamino ethylamine, usaf am-2, 1,2-ethanediamine, n,n-bis 1-methylethyl, n,n-diisopropyl-1,2-ethanediamine, n1,n1-diisopropylethane-1,2-diamine, 2-aminoethyl diisopropylamine, n,n-diisopropyl ethylenediamine, 2-diisopropylaminoethylamine
N',N'-di(propan-2-yl)ethane-1,2-diamine

Specifications

121-05-1
170°C
MFCD00015015
2734
CURJNMSGPBXOGK-UHFFFAOYSA-N
N',N'-di(propan-2-yl)ethane-1,2-diamine
8459
≥97.0% (GC,T)
N,N-Diisopropylethylenediamine
Yellow
C8H20N2
25 mL
2-aminoethyldiisopropylamine, n,n-diisopropylethylenediamine, 2-diisopropylamino ethylamine, usaf am-2, 1,2-ethanediamine, n,n-bis 1-methylethyl, n,n-diisopropyl-1,2-ethanediamine, n1,n1-diisopropylethane-1,2-diamine, 2-aminoethyl diisopropylamine, n,n-diisopropyl ethylenediamine, 2-diisopropylaminoethylamine
CC(C)N(CCN)C(C)C
144.262
144.26
Liquid
Videos
Safety and Handling

Safety and Handling

RTECSNumber : KV4200000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'