missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Dimethyl-1,3-propanediamine 99.0+%, TCI America™

Supplier:  TCI America D079025ML

 View more versions of this product

Catalog No. D079025ML


Explore available promotions

Description

Description

Specifications

Specifications

N,N-Dimethyl-1,3-propanediamine
Yellow
C5H14N2
25 mL
3-dimethylaminopropylamine, 3-dimethylamino propylamine, 3-dimethylamino-1-propylamine, n,n-dimethyl-1,3-propanediamine, 1,3-propanediamine, n,n-dimethyl, n,n-dimethyltrimethylenediamine, 3-aminopropyldimethylamine, n,n-dimethylpropane-1,3-diamine, 3-aminopropyl dimethylamine, dimethylaminopropylamine
CN(C)CCCN
102.181
102.18
Liquid
109-55-7
136°C
MFCD00008216
2734
IUNMPGNGSSIWFP-UHFFFAOYSA-N
N',N'-dimethylpropane-1,3-diamine
7993
≥99.0% (GC,T)

Chemical Identifiers

109-55-7
102.181
IUNMPGNGSSIWFP-UHFFFAOYSA-N
7993
CN(C)CCCN
C5H14N2
MFCD00008216
3-dimethylaminopropylamine, 3-dimethylamino propylamine, 3-dimethylamino-1-propylamine, n,n-dimethyl-1,3-propanediamine, 1,3-propanediamine, n,n-dimethyl, n,n-dimethyltrimethylenediamine, 3-aminopropyldimethylamine, n,n-dimethylpropane-1,3-diamine, 3-aminopropyl dimethylamine, dimethylaminopropylamine
N',N'-dimethylpropane-1,3-diamine
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0158

RTECSNumber : TX7525000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions