Learn More
N,N-Dimethyl-m-phenylenediamine dihydrochloride, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0040306
Description
it can be used to produce N-(4,5,6-trimethoxy-2,4-dimethyl-tetrahydro-pyran-3-yl)-formamide with 4,5,6-trimethoxy-2,4-dimethyl-tetrahydro-pyran-3-ylamine. This reaction will need solvent tetrahydrofuran. it is applied as a selective formylating agent for one of two amino groups in such basic amino acids as ornithine or lysine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsit can be used to produce N-(4,5,6-trimethoxy-2,4-dimethyl-tetrahydro-pyran-3-yl)-formamide with 4,5,6-trimethoxy-2,4-dimethyl-tetrahydro-pyran-3-ylamine. This reaction will need solvent tetrahydrofuran. it is applied as a selective formylating agent for one of two amino groups in such basic amino acids as ornithine or lysine.
Solubility
Soluble in water, chloroform and dichloromethane. Slightly soluble in ethyl acetate.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
N,N-Dimethyl-m-phenylenediamine dihydrochloride | |
216°C (decomposition) | |
MFCD00012971 | |
UN2811 | |
n1,n1-dimethylbenzene-1,3-diamine dihydrochloride, n,n-dimethyl-m-phenylenediamine dihydrochloride, m-dimethylamino aniline, n,n-dimethyl-1,3-phenylenediamine dihydrochloride, 1,3-benzenediamine, n,n-dimethyl-, dihydrochloride, n,n-dimethylbenzene-1,3-diamine dihydrochloride, 3-amino-n,n-dimethylaniline, 1,3-benzenediamine, n1,n1-dimethyl-, hydrochloride 1:2, pubchem14991, acmc-20a5k9 | |
BZJPIQKDEGXVFG-UHFFFAOYSA-N | |
3-N,3-N-dimethylbenzene-1,3-diamine;dihydrochloride | |
77124 | |
99% |
3575-32-4 | |
C8H14Cl2N2 | |
5 g | |
3694384 | |
Soluble in water,chloroform and dichloromethane. Slightly soluble in ethyl acetate. | |
CN(C)C1=CC=CC(=C1)N.Cl.Cl | |
209.114 | |
209.12 |
Chemical Identifiers
3575-32-4 | |
209.114 | |
BZJPIQKDEGXVFG-UHFFFAOYSA-N | |
77124 | |
CN(C)C1=CC=CC(=C1)N.Cl.Cl |
C8H14Cl2N2 | |
MFCD00012971 | |
n1,n1-dimethylbenzene-1,3-diamine dihydrochloride, n,n-dimethyl-m-phenylenediamine dihydrochloride, m-dimethylamino aniline, n,n-dimethyl-1,3-phenylenediamine dihydrochloride, 1,3-benzenediamine, n,n-dimethyl-, dihydrochloride, n,n-dimethylbenzene-1,3-diamine dihydrochloride, 3-amino-n,n-dimethylaniline, 1,3-benzenediamine, n1,n1-dimethyl-, hydrochloride 1:2, pubchem14991, acmc-20a5k9 | |
3-N,3-N-dimethylbenzene-1,3-diamine;dihydrochloride |
Safety and Handling
GHS H Statement
H300-H310-H330-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P333+P313-P361-P363-P501c
H301+H311+H331-H315-H317-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 222-693-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only