missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Dimethyl-m-phenylenediamine dihydrochloride, 99%, Thermo Scientific Chemicals

$119.84 - $448.70

Chemical Identifiers

CAS 3575-32-4
Molecular Formula C8H12N2·2HCl
Molecular Weight (g/mol) 209.12
MDL Number MFCD00012971
InChI Key BZJPIQKDEGXVFG-UHFFFAOYSA-N
Synonym n1,n1-dimethylbenzene-1,3-diamine dihydrochloride, n,n-dimethyl-m-phenylenediamine dihydrochloride, m-dimethylamino aniline, n,n-dimethyl-1,3-phenylenediamine dihydrochloride, 1,3-benzenediamine, n,n-dimethyl-, dihydrochloride, n,n-dimethylbenzene-1,3-diamine dihydrochloride, 3-amino-n,n-dimethylaniline, 1,3-benzenediamine, n1,n1-dimethyl-, hydrochloride 1:2, pubchem14991, acmc-20a5k9
PubChem CID 77124
IUPAC Name 3-N,3-N-dimethylbenzene-1,3-diamine;dihydrochloride
SMILES CN(C)C1=CC=CC(=C1)N.Cl.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC204340050
SDS View Documents
Thermo Scientific Chemicals
204340050
5 g Glass bottle
Each for $119.84
 
AC204340250
SDS View Documents
Thermo Scientific Chemicals
204340250
25 g Glass bottle
Each for $448.70
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3575-32-4
209.12
BZJPIQKDEGXVFG-UHFFFAOYSA-N
77124
CN(C)C1=CC=CC(=C1)N.Cl.Cl
C8H12N2·2HCl
MFCD00012971
n1,n1-dimethylbenzene-1,3-diamine dihydrochloride, n,n-dimethyl-m-phenylenediamine dihydrochloride, m-dimethylamino aniline, n,n-dimethyl-1,3-phenylenediamine dihydrochloride, 1,3-benzenediamine, n,n-dimethyl-, dihydrochloride, n,n-dimethylbenzene-1,3-diamine dihydrochloride, 3-amino-n,n-dimethylaniline, 1,3-benzenediamine, n1,n1-dimethyl-, hydrochloride 1:2, pubchem14991, acmc-20a5k9
3-N,3-N-dimethylbenzene-1,3-diamine;dihydrochloride

Specifications

3575-32-4
100.0
Purple to White
98.5% min. (Argentometry)
C8H12N2·2HCl
MFCD00012971
13, 40
Solubility in water: soluble
CN(C)C1=CC=CC(=C1)N.Cl.Cl
209.12
209.12
Crystalline Powder
98.5
217.0°C
Authentic
Glass bottle
(CH3)2NC6H4NH2·2HCl
5 g
n1,n1-dimethylbenzene-1,3-diamine dihydrochloride, n,n-dimethyl-m-phenylenediamine dihydrochloride, m-dimethylamino aniline, n,n-dimethyl-1,3-phenylenediamine dihydrochloride, 1,3-benzenediamine, n,n-dimethyl-, dihydrochloride, n,n-dimethylbenzene-1,3-diamine dihydrochloride, 3-amino-n,n-dimethylaniline, 1,3-benzenediamine, n1,n1-dimethyl-, hydrochloride 1:2, pubchem14991, acmc-20a5k9
BZJPIQKDEGXVFG-UHFFFAOYSA-N
3-N,3-N-dimethylbenzene-1,3-diamine;dihydrochloride
77124
99%
N, N-Dimethyl-m-phenylenediamine dihydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Toxic if inhaled.
Causes serious eye irritation.
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 222-693-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only