missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N,N-Dimethyl-N-(methylsulfanylmethylene)ammonium Iodide 98.0+%, TCI America™
$177.75 - $600.40
Chemical Identifiers
CAS | 29085-13-0 |
---|---|
Molecular Formula | C4H10INS |
Molecular Weight (g/mol) | 231.10 |
MDL Number | MFCD17015287 |
InChI Key | BPBAGFOLLWBKGJ-UHFFFAOYSA-M |
Synonym | N,N-Dimethyl-N-(methylsulfanylmethylene)iminium Iodide |
PubChem CID | 12347574 |
IUPAC Name | dimethyl[(methylsulfanyl)methylidene]azanium iodide |
SMILES | [I-].CSC=[N+](C)C |
Chemical Identifiers
29085-13-0 | |
231.10 | |
BPBAGFOLLWBKGJ-UHFFFAOYSA-M | |
12347574 | |
[I-].CSC=[N+](C)C |
C4H10INS | |
MFCD17015287 | |
N,N-Dimethyl-N-(methylsulfanylmethylene)iminium Iodide | |
dimethyl[(methylsulfanyl)methylidene]azanium iodide |
Specifications
29085-13-0 | |
White | |
MFCD17015287 | |
N,N-Dimethyl-N-(methylsulfanylmethylene)iminium Iodide | |
[I-].CSC=[N+](C)C | |
231.10 | |
231.10 | |
Crystalline Powder |
125°C | |
C4H10INS | |
5 g | |
BPBAGFOLLWBKGJ-UHFFFAOYSA-M | |
dimethyl[(methylsulfanyl)methylidene]azanium iodide | |
12347574 | |
≥98.0% (T) | |
N,N-Dimethyl-N-(methylsulfanylmethylene)ammonium Iodide |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator