missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dimethyl-N,N'-bis(3-methylaminopropyl)trimethylenediamine, tech. 90%, Thermo Scientific Chemicals

$224.19 - $224.19

Chemical Identifiers

CAS 123-67-1
Molecular Formula C13H32N4
Molecular Weight (g/mol) 244.427
MDL Number MFCD00014858
InChI Key XRTKMBBIDHUQNP-UHFFFAOYSA-N
Synonym 6,10-dimethyl-2,6,10,14-tetraazapentadecane, 2,6,10,14-tetraazapentadecane, 6,10-dimethyl, n1,n1'-propane-1,3-diyl bis n1,n3-dimethylpropane-1,3-diamine, n,n'-dimethyl-n,n'-bis 3-methylaminopropyl trimethylenediamine, 1,3-propanediamine, n,n'-dimethyl-n,n'-bis 3-methylamino propyl, n,n'-dimethyl-n,n'-bis 3-methylaminopropyl trimeth, 1,3-propanediamine, n1,n3-dimethyl-n1,n3-bis 3-methylamino propyl, ksc496s7p
PubChem CID 67160
IUPAC Name N,N'-dimethyl-N'-[3-[methyl-[3-(methylamino)propyl]amino]propyl]propane-1,3-diamine
SMILES CNCCCN(C)CCCN(C)CCCNC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6575803
View Documents
Thermo Scientific Chemicals
H6575803
1 g
Each for $224.19
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

123-67-1
244.427
XRTKMBBIDHUQNP-UHFFFAOYSA-N
67160
CNCCCN(C)CCCN(C)CCCNC
C13H32N4
MFCD00014858
6,10-dimethyl-2,6,10,14-tetraazapentadecane, 2,6,10,14-tetraazapentadecane, 6,10-dimethyl, n1,n1'-propane-1,3-diyl bis n1,n3-dimethylpropane-1,3-diamine, n,n'-dimethyl-n,n'-bis 3-methylaminopropyl trimethylenediamine, 1,3-propanediamine, n,n'-dimethyl-n,n'-bis 3-methylamino propyl, n,n'-dimethyl-n,n'-bis 3-methylaminopropyl trimeth, 1,3-propanediamine, n1,n3-dimethyl-n1,n3-bis 3-methylamino propyl, ksc496s7p
N,N'-dimethyl-N'-[3-[methyl-[3-(methylamino)propyl]amino]propyl]propane-1,3-diamine

Specifications

123-67-1
C13H32N4
1 g
6,10-dimethyl-2,6,10,14-tetraazapentadecane, 2,6,10,14-tetraazapentadecane, 6,10-dimethyl, n1,n1'-propane-1,3-diyl bis n1,n3-dimethylpropane-1,3-diamine, n,n'-dimethyl-n,n'-bis 3-methylaminopropyl trimethylenediamine, 1,3-propanediamine, n,n'-dimethyl-n,n'-bis 3-methylamino propyl, n,n'-dimethyl-n,n'-bis 3-methylaminopropyl trimeth, 1,3-propanediamine, n1,n3-dimethyl-n1,n3-bis 3-methylamino propyl, ksc496s7p
CNCCCN(C)CCCN(C)CCCNC
244.427
244.43
Technical
N,N'-Dimethyl-N,N'-bis(3-methylaminopropyl)trimethylenediamine
Yellow
MFCD00014858
3602172
XRTKMBBIDHUQNP-UHFFFAOYSA-N
N,N'-dimethyl-N'-[3-[methyl-[3-(methylamino)propyl]amino]propyl]propane-1,3-diamine
67160
90%
Liquid
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 204-641-9

TSCA : Yes

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only