missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N,N-Dimethyl-p-toluidine 98.0+%, TCI America™
Supplier: TCI America D0807500ML
Specifications
N,N-Dimethyl-p-toluidine | |
Yellow | |
C9H13N | |
500 mL | |
n,n-dimethyl-p-toluidine, dimethyl-p-toluidine, benzenamine, n,n,4-trimethyl, n,n-dimethyl-4-methylaniline, n,n-dimethyl-para-toluidine, 4,n,n-trimethylaniline, dimethyl-4-toluidine, p-dimethylamino toluene, n,n-dimethyl-p-tolylamine, n,n,4-trimethylbenzenamine | |
CN(C)C1=CC=C(C)C=C1 | |
135.21 | |
135.21 | |
Liquid |
99-97-8 | |
211°C | |
MFCD00008316 | |
2810 | |
GYVGXEWAOAAJEU-UHFFFAOYSA-N | |
N,N,4-trimethylaniline | |
7471 | |
≥98.0% (GC,T) |
Chemical Identifiers
99-97-8 | |
135.21 | |
GYVGXEWAOAAJEU-UHFFFAOYSA-N | |
7471 | |
CN(C)C1=CC=C(C)C=C1 |
C9H13N | |
MFCD00008316 | |
n,n-dimethyl-p-toluidine, dimethyl-p-toluidine, benzenamine, n,n,4-trimethyl, n,n-dimethyl-4-methylaniline, n,n-dimethyl-para-toluidine, 4,n,n-trimethylaniline, dimethyl-4-toluidine, p-dimethylamino toluene, n,n-dimethyl-p-tolylamine, n,n,4-trimethylbenzenamine | |
N,N,4-trimethylaniline |
Safety and Handling
EINECSNumber : (3)-0191
RTECSNumber : XU5803000
TSCA : Yes