missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Dimethylacetamide, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 375231000

 View more versions of this product

Catalog No. AC375231000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N, N-Dimethylacetamide
127-19-5
100.0
Colorless
164.0°C to 166.0°C
Authentic
99.4% min. (GC)
C4H9NO
CH3CON(CH3)2
100 mL
15, 3248
Solubility in water: soluble. Other solubilities: soluble in acetone,diethyl ether,benzene,,chloroform,ethanol and toluene
CN(C)C(C)=O
87.12
1.02 mPa.s (20°C)
87.12
Extra Dry over Molecular Sieve
Over Molecular Sieves
99.4
-20.0°C
0.9370g/mL
70°C
Water <50ppm
AcroSeal™ Glass Bottle
1.4370 to 1.4390
MFCD00008686
0.937
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
FXHOOIRPVKKKFG-UHFFFAOYSA-N
N,N-dimethylacetamide
31374
CHEBI:84254
99.5%
Liquid

Chemical Identifiers

127-19-5
87.12
FXHOOIRPVKKKFG-UHFFFAOYSA-N
31374
N,N-dimethylacetamide
C4H9NO
MFCD00008686
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
CHEBI:84254
CN(C)C(C)=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May damage the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and kee

GHS Signal Word: Danger

EINECSNumber : 204-826-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only