missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N,N-Dimethylacetamide, 99.9+%, OmniSolv™, MilliporeSigma™
Supplier: Technidata DX15436
Specifications
OmniPure™ | |
1 L | |
99.9+% | |
CH3CON(CH3)2 | |
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine | |
CN(C)C(C)=O | |
87.12 | |
CHEBI:84254 |
127-19-5 | |
Liquid | |
C4H9NO | |
MFCD00008686 | |
FXHOOIRPVKKKFG-UHFFFAOYSA-N | |
N,N-dimethylacetamide | |
31374 | |
87.12 |
Chemical Identifiers
127-19-5 | |
87.12 | |
FXHOOIRPVKKKFG-UHFFFAOYSA-N | |
31374 | |
N,N-dimethylacetamide |
C4H9NO | |
MFCD00008686 | |
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine | |
CHEBI:84254 | |
CN(C)C(C)=O |
Safety and Handling
Recommended Storage : Room Temperature