Learn More
N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
$129.91 - $2401.39
Chemical Identifiers
CAS | 127-19-5 |
---|---|
Molecular Formula | C4H9NO |
Molecular Weight (g/mol) | 87.12 |
MDL Number | MFCD00008686 |
InChI Key | FXHOOIRPVKKKFG-UHFFFAOYSA-N |
Synonym | dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine |
PubChem CID | 31374 |
ChEBI | CHEBI:84254 |
IUPAC Name | N,N-dimethylacetamide |
SMILES | CN(C)C(C)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC181070010
|
Thermo Scientific Chemicals
181070010 |
1 L | Glass Bottle |
Each for $129.91
Case of 4 Each for $0.00
|
|
|||||
AC181070025
|
Thermo Scientific Chemicals
181070025 |
2.5 L | Glass Bottle |
Each for $237.98
Case of 4 Each for $0.00
|
|
|||||
AC181070050
|
Thermo Scientific Chemicals
181070050 |
5 L | Plastic Drum |
Each for $435.30
|
|
|||||
AC181070250
|
Thermo Scientific Chemicals
181070250 |
25 L | Metal Drum |
Each for $2,401.39
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
127-19-5 | |
87.12 | |
FXHOOIRPVKKKFG-UHFFFAOYSA-N | |
31374 | |
N,N-dimethylacetamide |
C4H9NO | |
MFCD00008686 | |
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine | |
CHEBI:84254 | |
CN(C)C(C)=O |
Specifications
127-19-5 | |
100.0 | |
1 L | |
Glass Bottle | |
1.4370 to 1.4390 (20°C, 589nm) | |
99+% | |
CH3CON(CH3)2 | |
Vapor Pressure: 1.7mbar at 25°C | |
04, 59 | |
15, 3248 | |
FXHOOIRPVKKKFG-UHFFFAOYSA-N | |
N,N-dimethylacetamide | |
31374 | |
87.12 | |
Extra Pure |
99.0 | |
0.9370g/mL | |
Authentic | |
Liquid | |
0.01% max. | |
C4H9NO | |
MFCD00008686 | |
0.1% max. | |
01,270; 02,144; 04,165 | |
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine | |
CN(C)C(C)=O | |
87.12 | |
CHEBI:84254 | |
≥99.0% | |
N, N-Dimethylacetamide |
Safety and Handling
GHS P Statement
May damage the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Emergency Overview
Combustible liquid. Harmful in contact with skin. Harmful by inhalation. Irritating to eyes. May cause harm to the unborn child.
NFPA
Health:2 Flammability:2 Instability:0
Signal Word: WARNING!
RUO – Research Use Only