missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Dimethylacetamide, 99+%, for spectroscopy, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 167780025

Catalog No. AC167780025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

For UV/VIS spectroscopy
Specifications

Specifications

N, N-Dimethylacetamide
0.01 max. at 360nm, 0.03 max. at 320nm, 0.05 max. at 310nm, 0.15 max. at 290nm, 0.3 max. at 280nm, 1 max. at 270nm
100.0
164.0°C to 166.0°C
70°C
1.4370 to 1.4390
99+%
C4H9NO
MFCD00008686
0.05% max.
01,270; 02,144; 04,165
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
CN(C)C(C)=O
87.12
CHEBI:84254
≥99%
127-19-5
99.0
0.9370g/mL
2.5L
Glass bottle
0.0005% max.
0.937
CH3CON(CH3)2
1.02 mPa.s (20°C)
04, 59
15, 3248
FXHOOIRPVKKKFG-UHFFFAOYSA-N
N,N-dimethylacetamide
31374
87.12
Spectroscopy
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS P Statement
May damage the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only