missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacrylamide (stabilized with MEHQ) 99.0+%, TCI America™

$50.33 - $141.67

Chemical Identifiers

CAS 2680-03-7
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00008626
InChI Key YLGYACDQVQQZSW-UHFFFAOYSA-N
Synonym n,n-dimethylacrylamide, 2-propenamide, n,n-dimethyl, acrylamide, n,n-dimethyl, acylamide, n,n-dimethyl, n,n-dimethyl-acrylamide, n,n-dimethyl-2-propenamide, dimethylamid kyseliny akrylove, nn-dimethylacrylamide, unii-as46jk7q6i, propenamide, n,n-dimethyl
PubChem CID 17587
IUPAC Name N,N-dimethylprop-2-enamide
SMILES CN(C)C(=O)C=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D109125G
View Documents
TCI America
D109125G
25 g N/A
Only null left
 
D1091500G
View Documents
TCI America
D1091500G
500 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2680-03-7
99.13
YLGYACDQVQQZSW-UHFFFAOYSA-N
17587
CN(C)C(=O)C=C
C5H9NO
MFCD00008626
n,n-dimethylacrylamide, 2-propenamide, n,n-dimethyl, acrylamide, n,n-dimethyl, acylamide, n,n-dimethyl, n,n-dimethyl-acrylamide, n,n-dimethyl-2-propenamide, dimethylamid kyseliny akrylove, nn-dimethylacrylamide, unii-as46jk7q6i, propenamide, n,n-dimethyl
N,N-dimethylprop-2-enamide

Specifications

2680-03-7
80°C
MFCD00008626
2810
YLGYACDQVQQZSW-UHFFFAOYSA-N
N,N-dimethylprop-2-enamide
17587
≥99.0% (GC)
N,N-Dimethylacrylamide (stabilized with MEHQ)
Yellow
C5H9NO
25 g
n,n-dimethylacrylamide, 2-propenamide, n,n-dimethyl, acrylamide, n,n-dimethyl, acylamide, n,n-dimethyl, n,n-dimethyl-acrylamide, n,n-dimethyl-2-propenamide, dimethylamid kyseliny akrylove, nn-dimethylacrylamide, unii-as46jk7q6i, propenamide, n,n-dimethyl
CN(C)C(=O)C=C
99.13
99.13
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1017

RTECSNumber : AU3230000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'