Learn More
N,N-Dimethylethylenediamine, 97%, Thermo Scientific Chemicals
Alfa Aesar N,N-Dimethylethylenediamine, 97%, Quantity: 250g, Boiling Point: 107 deg.C, Melting Point: <-70 deg.C, Molecular Weight: 88.154, Percent Purity: 97%, Beilstein: 605279, CAS: 108-00-9, Density: 0.81, DOT Information: Hazard Class: 8; Packaging Group: I, EINECS Number: 203-541-2
Supplier: Thermo Scientific Chemicals B2018230
Description
It finds its application as an intermediate in the synthesis of Cefotiam. It is also used as a catalyst in the synthesis of allylic nitro compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds its application as an intermediate in the synthesis of Cefotiam. It is also used as a catalyst in the synthesis of allylic nitro compounds.
Solubility
Fully miscible in water.
Notes
Air sensitive & hygroscopic. Store away from oxidizing agents, air and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from electrostatic charges.
Specifications
N,N-Dimethylethylenediamine | |
<-70°C | |
107°C | |
C4H12N2 | |
(CH3)2NCH2CH2NH2 | |
250 g | |
605279 | |
n,n-dimethylethylenediamine, 2-dimethylaminoethylamine, 2-aminoethyl dimethylamine, 1,2-ethanediamine, n,n-dimethyl, 2-dimethylamino ethylamine, n,n-dimethyl-1,2-ethanediamine, n1,n1-dimethylethane-1,2-diamine, n,n-dimethyl-1,2-ethylenediamine, 2-aminoethyldimethylamine, n,n-dimethylethanediamine | |
DILRJUIACXKSQE-UHFFFAOYSA-N | |
N',N'-dimethylethane-1,2-diamine | |
66053 | |
97% |
108-00-9 | |
0.81 | |
12°C (53°F) | |
1.427 | |
MFCD00008175 | |
UN2733 | |
Air sensitive; Hygroscopic | |
Fully miscible in water. | |
CN(C)CCN | |
88.154 | |
88.15 |
Chemical Identifiers
108-00-9 | |
88.154 | |
DILRJUIACXKSQE-UHFFFAOYSA-N | |
66053 | |
CN(C)CCN |
C4H12N2 | |
MFCD00008175 | |
n,n-dimethylethylenediamine, 2-dimethylaminoethylamine, 2-aminoethyl dimethylamine, 1,2-ethanediamine, n,n-dimethyl, 2-dimethylamino ethylamine, n,n-dimethyl-1,2-ethanediamine, n1,n1-dimethylethane-1,2-diamine, n,n-dimethyl-1,2-ethylenediamine, 2-aminoethyldimethylamine, n,n-dimethylethanediamine | |
N',N'-dimethylethane-1,2-diamine |
Safety and Handling
GHS H Statement
H225-H314-H302-H312
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P363-P372-P374-P380-P501c
H225-H302+H312-H314
DOTInformation : Hazard Class: 8; Packaging Group: I
EINECSNumber : 203-541-2
RTECSNumber : KH8608250
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only