missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Dimethylformamide, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals

N,N-Dimethylformamide, 99.8%, C3H7NO, CAS Number-68-12-2, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dwumetyloformamid, dimethylformamide, dmfa, dimethyl formamide, dimetilformamide, dmf, dimethylformamid, 1L, 0.01 % max., Vapor Pressure: 4.9mbar at 20 deg.C, 04, 58

Supplier:  Thermo Scientific Chemicals 348431000

 View more versions of this product

Catalog No. AC348431000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N,N-Dimethylformamide
water <50 ppm
White to Beige
100.0
0.9450g/mL
100 mL
Authentic
Crystalline Powder
99.8%
C3H7NO
MFCD00003284
0.8 mPa.s (20°C)
04, 58
15, 3267
Solubility in water: soluble. Other solubilities: miscible with most organic solvents
CN(C)C=O
73.10
CHEBI:17741
99.8%
-61.0°C
68-12-2
99.75
>350°C
153.0°C
58°C
AcroSeal™ Glass Bottle
1.4290 to 1.4320
0.945
HCON(CH3)2
Vapor Pressure: 4.9mbar at 20°C
0.005% Max. (Coulometric)
01,278; 02,153; 03,115; 04,184; 05,247; 07,124; 08,189; 09,182; 11,198; 12,203; 13,124; 14,148; 16,144
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
ZMXDDKWLCZADIW-UHFFFAOYSA-N
N,N-dimethylformamide
6228
73.09
Extra Dry over Molecular Sieve

Chemical Identifiers

68-12-2
73.10
ZMXDDKWLCZADIW-UHFFFAOYSA-N
6228
N,N-dimethylformamide
C3H7NO
MFCD00003284
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
CHEBI:17741
CN(C)C=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May damage the unborn child.
Flammable liquid and vapour.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only