Learn More
N,N-Dimethylformamide, anhydrous, amine free, 99.9%, Thermo Scientific Chemicals
Suitable For Nucleic Acids, Peptide Synthesis
$261.34 - $862.08
Chemical Identifiers
CAS | 68-12-2 |
---|---|
Molecular Formula | C3H7NO |
Molecular Weight (g/mol) | 73.10 |
MDL Number | MFCD00003284 |
InChI Key | ZMXDDKWLCZADIW-UHFFFAOYSA-N |
Synonym | dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa |
PubChem CID | 6228 |
ChEBI | CHEBI:17741 |
IUPAC Name | N,N-dimethylformamide |
SMILES | CN(C)C=O |
Description
Suitable for nucleic acids, peptide synthesisN, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
68-12-2 | |
73.10 | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
6228 | |
N,N-dimethylformamide |
C3H7NO | |
MFCD00003284 | |
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
CHEBI:17741 | |
CN(C)C=O |
Specifications
68-12-2 | |
Colorless | |
153°C | |
440°C (824°F) | |
Amine-like | |
1.431 | |
4 L | |
605365 | |
Miscible with water and most organic solvents. | |
CN(C)C=O | |
73.10 | |
CHEBI:17741 | |
3mm Hg at 20°C | |
N, N-Dimethylformamide, Contains amine and water scavenging sieve |
-61°C | |
0.944 g/cm3 at 20°C | |
57°C (135°F) | |
99.9% | |
C3H7NO | |
MFCD00003284 | |
2265 | |
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
N,N-dimethylformamide | |
6228 | |
73.09 | |
Liquid |
Safety and Handling
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P312-P363-P370+P378q-P501c
H226-H312+H332-H319-H335-H336-H350-H360D
DOTInformation : DOT Class: 3, Packaging Group: III
EINECSNumber : 200-679-5
RTECSNumber : LQ2100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only