missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N,N-Dimethylformamide (Certified ACS), Fisher Chemical™
Meet or exceeds the latest ACS specifications. Actual lot of analysis on label
Supplier: Fisher Chemical D11920
Specifications
6 to 8 | |
-61°C | |
Colorless | |
0.945g/cm³ | |
20 L | |
Pass Test | |
Liquid | |
≥99.8 % | |
C3H7NO | |
MFCD00003284 | |
0.8 mPa/s at 20°C | |
0.15% max. | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
N,N-dimethylformamide | |
6228 | |
73.09 | |
Certified ACS Reagent |
N, N-Dimethylformamide | |
68-12-2 | |
>350°C | |
153°C | |
58°C (136.4°F) | |
Poly Pail | |
0.005% max. | |
0.0005mEq/g max. | |
0.003mEq/g max. | |
4.9mbar at 20°C | |
2265 | |
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
CN(C)C=O | |
73.10 | |
CHEBI:17741 | |
≥99.8% |
Chemical Identifiers
68-12-2 | |
73.10 | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
6228 | |
N,N-dimethylformamide |
C3H7NO | |
MFCD00003284 | |
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
CHEBI:17741 | |
CN(C)C=O |
Safety and Handling
WARNING!
Emergency Overview
Flammable liquid and vapor. May cause methemoglobinemia. Irritating to eyes and skin. Lachrymator (substance which increases the flow of tears). May cause central nervous system effects. May cause adverse liver effects. May cause adverse kidney effects. May cause harm to the unborn child. Use personal protective equipment. Keep away from open flames
DOTInformation : DOT Class 3, : Flammable Liquid