Learn More
N,N-Dimethylformamide di-tert-butyl Acetal, tech. 90%, Thermo Scientific Chemicals
$206.36 - $814.48
Chemical Identifiers
CAS | 36805-97-7 |
---|---|
Molecular Formula | C11H25NO2 |
Molecular Weight (g/mol) | 203.326 |
MDL Number | MFCD00015002 |
InChI Key | DBNQIOANXZVWIP-UHFFFAOYSA-N |
Synonym | n,n-dimethylformamide di-tert-butyl acetal, 1,1-di-tert-butoxytrimethylamine, 1,1-di-tert-butoxy-n,n-dimethylmethanamine, bis tert-butoxy methyl dimethylamine, 1,1-bis 1,1-dimethylethoxy-n,n,n-trimethylamine, di-tert-butoxymethyldimethylamine, n,n-dimethylformamide di-t-butyl acetal, methanamine, 1,1-bis 1,1-dimethylethoxy-n,n-dimethyl, 1,1-ditert-butoxy-n,n-dimethylmethanamine, 1,1-di-tert-butoxy-n,n-dimethylmethylamine |
PubChem CID | 547712 |
IUPAC Name | N,N-dimethyl-1,1-bis[(2-methylpropan-2-yl)oxy]methanamine |
SMILES | CC(C)(C)OC(N(C)C)OC(C)(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0043106
|
Thermo Scientific Chemicals
L0043106 |
5 g |
N/A
|
|
||||||
AAL0043114
|
Thermo Scientific Chemicals
L0043114 |
25 g |
N/A
|
|
||||||
Description
N,N-Dimethylformamide di-tert-butyl acetal is used as a reagent in the synthesis of (S)-2-(1-tert-butoxycarbonyl-2-{4-[2-(5-methyl-2-phenyloxazol-4-yl)ethoxy]phenyl}ethylamino)benzoic acid methyl ester, 3-O-tert-butylmorphine, tert-butylesters of pyrrole and indolecarboxylic acids. Further, it is used in the preparation of indolizines by intermolecular cyclization of picolinium salts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N-Dimethylformamide di-tert-butyl acetal is used as a reagent in the synthesis of (S)-2-(1-tert-butoxycarbonyl-2-{4-[2-(5-methyl-2-phenyloxazol-4-yl)ethoxy]phenyl}ethylamino)benzoic acid methyl ester, 3-O-tert-butylmorphine, tert-butylesters of pyrrole and indolecarboxylic acids. Further, it is used in the preparation of indolizines by intermolecular cyclization of picolinium salts.
Solubility
Miscible with toluene and benzene.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents and acids.
Chemical Identifiers
36805-97-7 | |
203.326 | |
DBNQIOANXZVWIP-UHFFFAOYSA-N | |
547712 | |
CC(C)(C)OC(N(C)C)OC(C)(C)C |
C11H25NO2 | |
MFCD00015002 | |
n,n-dimethylformamide di-tert-butyl acetal, 1,1-di-tert-butoxytrimethylamine, 1,1-di-tert-butoxy-n,n-dimethylmethanamine, bis tert-butoxy methyl dimethylamine, 1,1-bis 1,1-dimethylethoxy-n,n,n-trimethylamine, di-tert-butoxymethyldimethylamine, n,n-dimethylformamide di-t-butyl acetal, methanamine, 1,1-bis 1,1-dimethylethoxy-n,n-dimethyl, 1,1-ditert-butoxy-n,n-dimethylmethanamine, 1,1-di-tert-butoxy-n,n-dimethylmethylamine | |
N,N-dimethyl-1,1-bis[(2-methylpropan-2-yl)oxy]methanamine |
Specifications
36805-97-7 | |
75°C to 77°C (12 mmHg) | |
C11H25NO2 | |
(CH3)2NCH[OC(CH3)3]2 | |
5 g | |
969629 | |
n,n-dimethylformamide di-tert-butyl acetal, 1,1-di-tert-butoxytrimethylamine, 1,1-di-tert-butoxy-n,n-dimethylmethanamine, bis tert-butoxy methyl dimethylamine, 1,1-bis 1,1-dimethylethoxy-n,n,n-trimethylamine, di-tert-butoxymethyldimethylamine, n,n-dimethylformamide di-t-butyl acetal, methanamine, 1,1-bis 1,1-dimethylethoxy-n,n-dimethyl, 1,1-ditert-butoxy-n,n-dimethylmethanamine, 1,1-di-tert-butoxy-n,n-dimethylmethylamine | |
DBNQIOANXZVWIP-UHFFFAOYSA-N | |
N,N-dimethyl-1,1-bis[(2-methylpropan-2-yl)oxy]methanamine | |
547712 | |
90% | |
N,N-Dimethylformamide di-tert-butyl acetal |
0.85 | |
33°C (91°F) | |
1.412 | |
MFCD00015002 | |
UN1993 | |
Moisture sensitive | |
Miscible with toluene and benzene. | |
CC(C)(C)OC(N(C)C)OC(C)(C)C | |
203.326 | |
203.33 | |
Technical |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 253-222-7
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only