Learn More
N,N-Dimethylformamide dimethyl acetal, ca. 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 161255000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentSpecifications
N, N-Dimethylformamide dimethyl acetal | |
4637-24-5,149-73-5,68-12-2,67-56-1 | |
100.0 | |
0.8900g/mL | |
7°C | |
96% min. (GC) | |
C5H13NO2 | |
(CH3)2NCH(OCH3)2 | |
500 g | |
03,115; 04,184; 05,254; 06,221; 08,191; 09,182; 10,158; 13,120; 05,646; 06,566; 07,355; 09,447; 11,501 | |
n,n-dimethylformamide dimethyl acetal, 1,1-dimethoxytrimethylamine, dimethoxymethyl dimethylamine, methanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylformamide-dimethylacetal, dmf dimethyl acetal, dmfdma, dimethylformamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylmethylamine, dimethoxy dimethylamino methane | |
ZSXGLVDWWRXATF-UHFFFAOYSA-N | |
(dimethoxymethyl)dimethylamine | |
78373 | |
119.16 | |
Liquid |
ca 97% | |
96.0 | |
Colorless | |
102.0°C to 104.0°C | |
Authentic | |
Glass bottle | |
1.3972 | |
MFCD00008482 | |
1993 | |
0.89 | |
Solubility in water: hydrolysis. Other solubilities: soluble in many organic solvents | |
COC(OC)N(C)C | |
119.16 | |
CHEBI:85061 | |
ca. 97% |
Chemical Identifiers
4637-24-5 | |
119.16 | |
ZSXGLVDWWRXATF-UHFFFAOYSA-N | |
78373 | |
(dimethoxymethyl)dimethylamine |
C5H13NO2 | |
MFCD00008482 | |
n,n-dimethylformamide dimethyl acetal, 1,1-dimethoxytrimethylamine, dimethoxymethyl dimethylamine, methanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylformamide-dimethylacetal, dmf dimethyl acetal, dmfdma, dimethylformamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylmethylamine, dimethoxy dimethylamino methane | |
CHEBI:85061 | |
COC(OC)N(C)C |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye damage.
May cause an allergic skin reaction.
Harmful if inhaled.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF exposed or concerned: Get medical attention/advice.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face p
GHS Signal Word: Danger
EINECSNumber : 225-063-3
TSCA : TSCA
RUO – Research Use Only