missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Dimethyloctadecylamine, 89%, tech., Thermo Scientific Chemicals

$174.66 - $331.77

Chemical Identifiers

CAS 124-28-7
Molecular Formula C20H44ClN
Molecular Weight (g/mol) 334.03
MDL Number MFCD00048496
InChI Key PFKRTWCFCOUBHS-UHFFFAOYSA-N
Synonym n,n-dimethyloctadecylamine, dymanthine, dimantine, n,n-dimethyl-n-octadecylamine, dimethyloctadecylamine, 1-octadecanamine, n,n-dimethyl, n,n-dimethyl-1-octadecanamine, dimethyl stearamine, stearyldimethylamine, n,n-dimethylstearylamine
PubChem CID 15365
IUPAC Name N,N-dimethyloctadecan-1-amine
SMILES [Cl-].CCCCCCCCCCCCCCCCCC[NH+](C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC408432500
SDS View Documents
Thermo Scientific Chemicals
408432500
250 g Glass bottle
Each for $174.66
 
AC408430010
SDS View Documents
Thermo Scientific Chemicals
408430010
1 kg Glass bottle
Each for $331.77
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

124-28-7
334.03
PFKRTWCFCOUBHS-UHFFFAOYSA-N
15365
[Cl-].CCCCCCCCCCCCCCCCCC[NH+](C)C
C20H44ClN
MFCD00048496
n,n-dimethyloctadecylamine, dymanthine, dimantine, n,n-dimethyl-n-octadecylamine, dimethyloctadecylamine, 1-octadecanamine, n,n-dimethyl, n,n-dimethyl-1-octadecanamine, dimethyl stearamine, stearyldimethylamine, n,n-dimethylstearylamine
N,N-dimethyloctadecan-1-amine

Specifications

124-28-7
100.0
Colorless to Yellow
347.0°C
Authentic
Glass bottle
(CH3)2N(CH2)17CH3
250 g
15, 3522
Solubility in water: slightly soluble
[Cl-].CCCCCCCCCCCCCCCCCC[NH+](C)C
334.03
297.57
Technical
N, N-Dimethyloctadecylamine, tech., 89%
84.0
15.0°C to 20.0°C
0.8000g/mL
155°C
84% min. (C18) (GC)
C20H44ClN
MFCD00048496
0.8
n,n-dimethyloctadecylamine, dymanthine, dimantine, n,n-dimethyl-n-octadecylamine, dimethyloctadecylamine, 1-octadecanamine, n,n-dimethyl, n,n-dimethyl-1-octadecanamine, dimethyl stearamine, stearyldimethylamine, n,n-dimethylstearylamine
PFKRTWCFCOUBHS-UHFFFAOYSA-N
N,N-dimethyloctadecan-1-amine
15365
89%
Liquid After Melting
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 204-694-8

RTECSNumber : RG4200000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only