Learn More
N,N-Dimethyloctylamine, 97%
CAS: 7378-99-6 | C10H23N | 157.30 g/mol
$148.72 - $320.42
Chemical Identifiers
| CAS | 7378-99-6 |
|---|---|
| Molecular Formula | C10H23N |
| Molecular Weight (g/mol) | 157.30 |
| MDL Number | MFCD00009558 |
| InChI Key | UQKAOOAFEFCDGT-UHFFFAOYSA-N |
| Synonym | n,n-dimethyloctylamine, n,n-dimethyl-n-octylamine, dimethyloctylamine, octyldimethylamine, dimethyl octyl amine, n-octyldimethylamine, dimethyl-n-octylamine, 1-octanamine, n,n-dimethyl, octylamine, n,n-dimethyl, unii-20n7h7x4sd |
| PubChem CID | 16224 |
| IUPAC Name | N,N-dimethyloctan-1-amine |
| SMILES | CCCCCCCCN(C)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC225290250
|
Thermo Scientific Chemicals
225290250 |
25 mL | Glass bottle |
Each for $148.72
|
|
||||
|
AC225295000
|
Thermo Scientific Chemicals
225295000 |
500 mL | Glass bottle |
Each for $320.42
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7378-99-6 | |
| 157.30 | |
| UQKAOOAFEFCDGT-UHFFFAOYSA-N | |
| 16224 | |
| CCCCCCCCN(C)C |
| C10H23N | |
| MFCD00009558 | |
| n,n-dimethyloctylamine, n,n-dimethyl-n-octylamine, dimethyloctylamine, octyldimethylamine, dimethyl octyl amine, n-octyldimethylamine, dimethyl-n-octylamine, 1-octanamine, n,n-dimethyl, octylamine, n,n-dimethyl, unii-20n7h7x4sd | |
| N,N-dimethyloctan-1-amine |
Specifications
| 7378-99-6 | |
| 100.0 | |
| 0.7650g/mL | |
| 65°C | |
| 96% min. (C8-chains) (GC) | |
| C10H23N | |
| CH3(CH2)7N(CH3)2 | |
| 25 mL | |
| 0.765 | |
| Solubility in water: slightly soluble | |
| CCCCCCCCN(C)C | |
| 157.30 | |
| 157.3 | |
| Liquid |
| 96.0 | |
| -57.0°C | |
| 195.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4233 to 1.4253 | |
| MFCD00009558 | |
| 04,I,386 | |
| n,n-dimethyloctylamine, n,n-dimethyl-n-octylamine, dimethyloctylamine, octyldimethylamine, dimethyl octyl amine, n-octyldimethylamine, dimethyl-n-octylamine, 1-octanamine, n,n-dimethyl, octylamine, n,n-dimethyl, unii-20n7h7x4sd | |
| UQKAOOAFEFCDGT-UHFFFAOYSA-N | |
| N,N-dimethyloctan-1-amine | |
| 16224 | |
| 97% | |
| N, N-Dimethyloctylamine, 97% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
Very toxic to aquatic life.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contac
GHS Signal Word: Danger
EINECSNumber : 230-939-3
RTECSNumber : RG8075000
TSCA : TSCA
RUO – Research Use Only