missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethyloxamic Acid 97.0+%, TCI America™
$59.65 - $154.71
Chemical Identifiers
| CAS | 32833-96-8 |
|---|---|
| Molecular Formula | C4H7NO3 |
| Molecular Weight (g/mol) | 117.104 |
| MDL Number | MFCD01971023 |
| InChI Key | YKFGLGXRUVEMNF-UHFFFAOYSA-N |
| Synonym | N,N-Dimethyloxamidic Acid |
| PubChem CID | 2760995 |
| IUPAC Name | 2-(dimethylamino)-2-oxoacetic acid |
| SMILES | CN(C)C(=O)C(=O)O |
Chemical Identifiers
| 32833-96-8 | |
| 117.104 | |
| YKFGLGXRUVEMNF-UHFFFAOYSA-N | |
| 2760995 | |
| CN(C)C(=O)C(=O)O |
| C4H7NO3 | |
| MFCD01971023 | |
| N,N-Dimethyloxamidic Acid | |
| 2-(dimethylamino)-2-oxoacetic acid |
Specifications
| 32833-96-8 | |
| White | |
| MFCD01971023 | |
| N,N-Dimethyloxamidic Acid | |
| CN(C)C(=O)C(=O)O | |
| 117.104 | |
| 117.10 | |
| Crystalline Powder |
| 125°C | |
| C4H7NO3 | |
| 1 g | |
| YKFGLGXRUVEMNF-UHFFFAOYSA-N | |
| 2-(dimethylamino)-2-oxoacetic acid | |
| 2760995 | |
| ≥97.0% (T) | |
| N,N-Dimethyloxamic Acid |
Safety and Handling
TSCA : No