Learn More
N,N-Dimethylphosphoramidodichloridate, 97%, Thermo Scientific Chemicals
$122.52 - $379.85
Chemical Identifiers
CAS | 677-43-0 |
---|---|
Molecular Formula | C2H6Cl2NOP |
Molecular Weight (g/mol) | 161.95 |
MDL Number | MFCD00013896 |
InChI Key | YNHXBEVSSILHPI-UHFFFAOYSA-N |
Synonym | dimethylphosphoramidic dichloride, n,n-dimethylphosphoramidodichloridate, dichlorophosphoric dimethylamide, phosphoramidic dichloride, dimethyl, dimethylamido phosphoric dichloride, dimethylamidophosphoric acid dichloride, dimethylaminophosphoryl dichloride, n,n-dimethylphosphoramidic dichloride, dimethylamidophosphoric dichloride, n,n-dimethylphosphoramic dichloride |
PubChem CID | 12673 |
IUPAC Name | N-dichlorophosphoryl-N-methylmethanamine |
SMILES | CN(C)P(=O)(Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0723106
|
Thermo Scientific Chemicals
L0723106 |
5 g |
Each for $122.52
|
|
||||||
AAL0723114
|
Thermo Scientific Chemicals
L0723114 |
25 g |
Each for $379.85
|
|
||||||
Description
N,N-Dimethylphosphoramidodichloridate is used in the synthesis of phosphoramidates as well as (RS)-ethyl N,N-dimethylphosphoramidocyanidate (Nerve agent GA) which is used as a chemical weapon. It forms active intermediates, which react with alcohols in situ to give esters. It is also used to convert primary alcohols to alkyl chlorides. Furthe, it serves as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. In addition to this, it reacts with 1,2 diols to prepare cyclic phosphoric amides followed by alkenes after reduction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N-Dimethylphosphoramidodichloridate is used in the synthesis of phosphoramidates as well as (RS)-ethyl N,N-dimethylphosphoramidocyanidate (Nerve agent GA) which is used as a chemical weapon. It forms active intermediates, which react with alcohols in situ to give esters. It is also used to convert primary alcohols to alkyl chlorides. Furthe, it serves as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. In addition to this, it reacts with 1,2 diols to prepare cyclic phosphoric amides followed by alkenes after reduction.
Solubility
Miscible with dimethoxyethane.
Notes
Moisture and air sensitive. Hygroscopic. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Chemical Identifiers
677-43-0 | |
161.95 | |
YNHXBEVSSILHPI-UHFFFAOYSA-N | |
12673 | |
CN(C)P(=O)(Cl)Cl |
C2H6Cl2NOP | |
MFCD00013896 | |
dimethylphosphoramidic dichloride, n,n-dimethylphosphoramidodichloridate, dichlorophosphoric dimethylamide, phosphoramidic dichloride, dimethyl, dimethylamido phosphoric dichloride, dimethylamidophosphoric acid dichloride, dimethylaminophosphoryl dichloride, n,n-dimethylphosphoramidic dichloride, dimethylamidophosphoric dichloride, n,n-dimethylphosphoramic dichloride | |
N-dichlorophosphoryl-N-methylmethanamine |
Specifications
677-43-0 | |
89°C to 90°C (20 mmHg) | |
C2H6Cl2NOP | |
MFCD00013896 | |
UN3265 | |
Moisture sensitive | |
Miscible with dimethoxyethane. | |
CN(C)P(=O)(Cl)Cl | |
161.95 | |
161.96 | |
N,N-Dimethylphosphoramidodichloridate |
1.36 | |
>110°C (230°F) | |
1.463 | |
5 g | |
969956 | |
dimethylphosphoramidic dichloride, n,n-dimethylphosphoramidodichloridate, dichlorophosphoric dimethylamide, phosphoramidic dichloride, dimethyl, dimethylamido phosphoric dichloride, dimethylamidophosphoric acid dichloride, dimethylaminophosphoryl dichloride, n,n-dimethylphosphoramidic dichloride, dimethylamidophosphoric dichloride, n,n-dimethylphosphoramic dichloride | |
YNHXBEVSSILHPI-UHFFFAOYSA-N | |
N-dichlorophosphoryl-N-methylmethanamine | |
12673 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 211-641-2
RTECSNumber : TB4025000
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only