missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N,N'-Methylenebisacrylamide, 96%, extra pure, Thermo Scientific Chemicals
$63.06 - $199.38
Chemical Identifiers
CAS | 110-26-9 |
---|---|
Molecular Formula | C7H10N2O2 |
Molecular Weight (g/mol) | 154.17 |
InChI Key | ZIUHHBKFKCYYJD-UHFFFAOYSA-N |
Synonym | n,n'-methylenebisacrylamide, n,n'-methylenediacrylamide, n,n'-methylene-bis-acrylamide, methylenebisacrylamide, methylenediacrylamide, 2-propenamide, n,n'-methylenebis, bis-acrylamide, n,n'-methylenebis acrylamide, n,n'-methylidenebisacrylamide, bisacrylamide |
PubChem CID | 8041 |
IUPAC Name | N-[(prop-2-enoylamino)methyl]prop-2-enamide |
SMILES | C=CC(=O)NCNC(=O)C=C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC163801000
|
Thermo Scientific Chemicals
163801000 |
100 g | Plastic bottle |
Each for $63.06
|
|
||||
AC163805000
|
Thermo Scientific Chemicals
163805000 |
500 g | Plastic bottle |
Each for $199.38
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-26-9 | |
154.17 | |
n,n'-methylenebisacrylamide, n,n'-methylenediacrylamide, n,n'-methylene-bis-acrylamide, methylenebisacrylamide, methylenediacrylamide, 2-propenamide, n,n'-methylenebis, bis-acrylamide, n,n'-methylenebis acrylamide, n,n'-methylidenebisacrylamide, bisacrylamide | |
N-[(prop-2-enoylamino)methyl]prop-2-enamide |
C7H10N2O2 | |
ZIUHHBKFKCYYJD-UHFFFAOYSA-N | |
8041 | |
C=CC(=O)NCNC(=O)C=C |
Specifications
110-26-9 | |
White | |
95% min. (HPLC) | |
C7H10N2O2 | |
100 g | |
Solubility in water: soluble. Other solubilities: soluble in ethanol | |
C=CC(=O)NCNC(=O)C=C | |
154.17 | |
154.17 | |
Extra Pure | |
N, N'-Methylenebisacrylamide |
185°C | |
Authentic | |
Plastic bottle | |
(H2C=CHCONH)2CH2 | |
n,n'-methylenebisacrylamide, n,n'-methylenediacrylamide, n,n'-methylene-bis-acrylamide, methylenebisacrylamide, methylenediacrylamide, 2-propenamide, n,n'-methylenebis, bis-acrylamide, n,n'-methylenebis acrylamide, n,n'-methylidenebisacrylamide, bisacrylamide | |
ZIUHHBKFKCYYJD-UHFFFAOYSA-N | |
N-[(prop-2-enoylamino)methyl]prop-2-enamide | |
8041 | |
96% | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
GHS Signal Word: Warning
EINECSNumber : 203-750-9
RUO – Research Use Only