Learn More
Thermo Scientific Chemicals N,N'-Methylenebisacrylamide, 99+%, Electrophoresis Grade
$91.52 - $447.97
Chemical Identifiers
CAS | 110-26-9 |
---|---|
Molecular Formula | C7H10N2O2 |
Molecular Weight (g/mol) | 154.169 |
MDL Number | MFCD00008625 |
InChI Key | ZIUHHBKFKCYYJD-UHFFFAOYSA-N |
Synonym | n,n'-methylenebisacrylamide, n,n'-methylenediacrylamide, n,n'-methylene-bis-acrylamide, methylenebisacrylamide, methylenediacrylamide, 2-propenamide, n,n'-methylenebis, bis-acrylamide, n,n'-methylenebis acrylamide, n,n'-methylidenebisacrylamide, bisacrylamide |
PubChem CID | 8041 |
IUPAC Name | N-[(prop-2-enoylamino)methyl]prop-2-enamide |
SMILES | C=CC(=O)NCNC(=O)C=C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6671014
|
Thermo Scientific Chemicals
J6671014 |
25 g |
Each for $91.52
|
|
|||||
AAJ6671022
|
Thermo Scientific Chemicals
J6671022 |
100 g |
Each for $206.10
|
|
|||||
AAJ6671030
|
Thermo Scientific Chemicals
J6671030 |
250 g |
Each for $447.97
|
|
|||||
Description
N,N'-Methylenebisacrylamide is used in the preparation of polyacrylamide gels for electrophoretic separation of nucleic acids and proteins. It has also been used as a test compound for studying the biochemical isolation of insoluble Tau in transgenic mouse models of tauopathy by examining the two main isolation methods sarkosyl and formic acid extraction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N′-Methylenebisacrylamide is used in the preparation of polyacrylamide gels for electrophoretic separation of nucleic acids and proteins. It has also been used as a test compound for studying the biochemical isolation of insoluble Tau in transgenic mouse models of tauopathy by examining the two main isolation methods sarkosyl and formic acid extraction.
Solubility
Soluble in water or methanol
Notes
Air & Light Sensitive. Store in the dark. Store under dry inert gas. Incompatible with light, heat, air and oxidizing agents. Store at 4°C.
Chemical Identifiers
110-26-9 | |
154.169 | |
ZIUHHBKFKCYYJD-UHFFFAOYSA-N | |
8041 | |
C=CC(=O)NCNC(=O)C=C |
C7H10N2O2 | |
MFCD00008625 | |
n,n'-methylenebisacrylamide, n,n'-methylenediacrylamide, n,n'-methylene-bis-acrylamide, methylenebisacrylamide, methylenediacrylamide, 2-propenamide, n,n'-methylenebis, bis-acrylamide, n,n'-methylenebis acrylamide, n,n'-methylidenebisacrylamide, bisacrylamide | |
N-[(prop-2-enoylamino)methyl]prop-2-enamide |
Specifications
110-26-9 | |
White | |
C7H10N2O2 | |
25 g | |
Air and light sensitive | |
Soluble in water or methanol | |
C=CC(=O)NCNC(=O)C=C | |
154.169 | |
154.17 | |
Electrophoresis | |
N,N'-Methylenebisacrylamide |
>300°C | |
1.235 g/mL | |
MFCD00008625 | |
1706297 | |
n,n'-methylenebisacrylamide, n,n'-methylenediacrylamide, n,n'-methylene-bis-acrylamide, methylenebisacrylamide, methylenediacrylamide, 2-propenamide, n,n'-methylenebis, bis-acrylamide, n,n'-methylenebis acrylamide, n,n'-methylidenebisacrylamide, bisacrylamide | |
ZIUHHBKFKCYYJD-UHFFFAOYSA-N | |
N-[(prop-2-enoylamino)methyl]prop-2-enamide | |
8041 | |
≥99% | |
Powder |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 203-750-9
RTECSNumber : AS3678000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only