missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L16280AE

 View more versions of this product

Catalog No. AAL16280AE


Explore available promotions

Description

Description

N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine is used as a catalyst in the production of urethane foams. It acts as a complexing agent, plasticizer, surfactant solubilizer and curing agent for epoxy resin. It is also used as a viscosity modifier and as an anti-bloating agent for cattle. It is utilized in piezoelectric crystal detectors of sulfur dioxide and in additives for paint. It serves as a biological buffer as well as reagent for manganese. Further, it finds application in adhesives and sealant chemicals. In addition to this, it is used as a cross-linking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N,N′,N′-Tetrakis(2-hydroxypropyl)ethylenediamine is used as a catalyst in the production of urethane foams. It acts as a complexing agent, plasticizer, surfactant solubilizer and curing agent for epoxy resin. It is also used as a viscosity modifier and as an anti-bloating agent for cattle. It is utilized in piezoelectric crystal detectors of sulfur dioxide and in additives for paint. It serves as a biological buffer as well as reagent for manganese. Further, it finds application in adhesives and sealant chemicals. In addition to this, it is used as a cross-linking agent.

Solubility
Miscible with water, methanol, ethanol, toluene, ethylene glycol and perchloroethylene.

Notes
Incompatible with acids, acid chlorides, acid anhydrides, oxidizing agents and chloroformates.
Specifications

Specifications

N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine
1.03
>110°C (230°F)
C14H32N2O4
MFCD00004534
14,3599
Miscible with water,methanol,ethanol,toluene,ethylene glycol and perchloroethylene.
CC(CN(CCN(CC(C)O)CC(C)O)CC(C)O)O
292.42
292.42
102-60-3
240°C (10 mmHg)
Odorless
1.48
100 mL
quadrol, edetol, n,n,n',n'-tetrakis 2-hydroxypropyl ethylenediamine, entprol, adeka quadrol, quadrol l, neutrol te, 1,1',1,1'-ethylenedinitrilo tetra-2-propanol, edetolum latin, 2-propanol, 1,1',1,1'-1,2-ethanediyldinitrilo tetrakis
NSOXQYCFHDMMGV-UHFFFAOYSA-N
1-[2-[bis(2-hydroxypropyl)amino]ethyl-(2-hydroxypropyl)amino]propan-2-ol
7615
99%

Chemical Identifiers

102-60-3
292.42
NSOXQYCFHDMMGV-UHFFFAOYSA-N
7615
CC(CN(CCN(CC(C)O)CC(C)O)CC(C)O)O
C14H32N2O4
MFCD00004534
quadrol, edetol, n,n,n',n'-tetrakis 2-hydroxypropyl ethylenediamine, entprol, adeka quadrol, quadrol l, neutrol te, 1,1',1,1'-ethylenedinitrilo tetra-2-propanol, edetolum latin, 2-propanol, 1,1',1,1'-1,2-ethanediyldinitrilo tetrakis
1-[2-[bis(2-hydroxypropyl)amino]ethyl-(2-hydroxypropyl)amino]propan-2-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H317
May cause an allergic skin reaction.

P261-P271-P272-P280g-P302+P352-P304+P340-P312-P333+P313-P363-P501c

H317-H335

EINECSNumber : 203-041-4

RTECSNumber : UB5604000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only