Learn More
N,N,N',N'-Tetramethyl-1,3-propanediamine, 99+%, Thermo Scientific Chemicals
$73.08 - $73.08
Chemical Identifiers
CAS | 110-95-2 |
---|---|
Molecular Formula | C7H18N2 |
Molecular Weight (g/mol) | 130.23 |
MDL Number | MFCD00008337 |
InChI Key | DMQSHEKGGUOYJS-UHFFFAOYSA-N |
Synonym | n,n,n',n'-tetramethyl-1,3-propanediamine, 1,3-bis dimethylamino propane, tetramethyltrimethylenediamine, n1,n1,n3,n3-tetramethylpropane-1,3-diamine, n,n,n',n'-tetramethyl-1,3-diaminopropane, 1,3-propanediamine, n,n,n',n'-tetramethyl, bis dimethylamino methyl methane, n,n,n',n'-tetramethyltrimethylenediamine, unii-73b9i6hn5r |
PubChem CID | 8084 |
IUPAC Name | N,N,N',N'-tetramethylpropane-1,3-diamine |
SMILES | CN(C)CCCN(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC155510500
|
Thermo Scientific Chemicals
155510500 |
50 g | Glass bottle |
Each for $73.08
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-95-2 | |
130.23 | |
DMQSHEKGGUOYJS-UHFFFAOYSA-N | |
8084 | |
CN(C)CCCN(C)C |
C7H18N2 | |
MFCD00008337 | |
n,n,n',n'-tetramethyl-1,3-propanediamine, 1,3-bis dimethylamino propane, tetramethyltrimethylenediamine, n1,n1,n3,n3-tetramethylpropane-1,3-diamine, n,n,n',n'-tetramethyl-1,3-diaminopropane, 1,3-propanediamine, n,n,n',n'-tetramethyl, bis dimethylamino methyl methane, n,n,n',n'-tetramethyltrimethylenediamine, unii-73b9i6hn5r | |
N,N,N',N'-tetramethylpropane-1,3-diamine |
Specifications
110-95-2 | |
100.0 | |
Colorless to Yellow | |
145.0°C to 146.0°C | |
Authentic | |
Glass bottle | |
1.4224 to 1.4244 | |
MFCD00008337 | |
04, 262 | |
n,n,n',n'-tetramethyl-1,3-propanediamine, 1,3-bis dimethylamino propane, tetramethyltrimethylenediamine, n1,n1,n3,n3-tetramethylpropane-1,3-diamine, n,n,n',n'-tetramethyl-1,3-diaminopropane, 1,3-propanediamine, n,n,n',n'-tetramethyl, bis dimethylamino methyl methane, n,n,n',n'-tetramethyltrimethylenediamine, unii-73b9i6hn5r | |
CN(C)CCCN(C)C | |
130.23 | |
130.23 | |
Liquid |
99.0 | |
-70.0°C | |
0.7800g/mL | |
31°C | |
99% min. (GC) | |
C7H18N2 | |
(CH3)2N(CH2)3N(CH3)2 | |
50 g | |
0.78 | |
DMQSHEKGGUOYJS-UHFFFAOYSA-N | |
N,N,N',N'-tetramethylpropane-1,3-diamine | |
8084 | |
99+% | |
N,N,N′,N′-Tetramethyl-1,3-propanediamine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Toxic if inhaled.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth
GHS Signal Word: Danger
EINECSNumber : 203-818-8
RUO – Research Use Only