missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1210718

 View more versions of this product

Catalog No. AAA1210718


Explore available promotions

Description

Description

N,N,N',N'-tetramethyl-p-phenylenediamine dihydrochloride (TMPD) is an oxidizable compound that serves as a reducing co-substrate for heme peroxidases. It is also used as a test reagent in microbiology for the classification of cytochrome oxidase positive aerobic microorganisms. It finds application as a detection of peroxidases on polyacrylamide gels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N,N′,N′-tetramethyl-p-phenylenediamine dihydrochloride (TMPD) is an oxidizable compound that serves as a reducing co-substrate for heme peroxidases. It is also used as a test reagent in microbiology for the classification of cytochrome oxidase positive aerobic microorganisms. It finds application as a detection of peroxidases on polyacrylamide gels.

Solubility
Soluble in dimethyl sulfoxide, water and phosphate buffered saline at pH 7.2.

Notes
Light sensitive. Incompatible with strong oxidizing agents.                    
Specifications

Specifications

N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride
∼228°C (decomposition)
C10H18Cl2N2
50 g
Light sensitive
Soluble in dimethyl sulfoxide,water and phosphate buffered saline at pH 7.2.
CN(C)C1=CC=C(C=C1)N(C)C.Cl.Cl
237.168
237.17
637-01-4
99%
MFCD00012482
5132096
n,n,n',n'-tetramethyl-p-phenylenediamine dihydrochloride, wurster's reagent dihydrochloride, n,n,n',n'-tetramethyl-1,4-phenylenediamine dihydrochloride, unii-66w8hka51x, n1,n1,n4,n4-tetramethylbenzene-1,4-diamine dihydrochloride, wurster's blue dihydrochloride, 1,4-benzenediamine, n,n,n',n'-tetramethyl-, dihydrochloride, 1,4-benzenediamine, n1,n1,n4,n4-tetramethyl-, hydrochloride 1:2, 1-n,1-n,4-n,4-n-tetramethylbenzene-1,4-diamine dihydrochloride, wursters reagent
FBHKTSXMTASXFJ-UHFFFAOYSA-N
1-N,1-N,4-N,4-N-tetramethylbenzene-1,4-diamine;dihydrochloride
71561
≥98%

Chemical Identifiers

637-01-4
237.168
FBHKTSXMTASXFJ-UHFFFAOYSA-N
71561
CN(C)C1=CC=C(C=C1)N(C)C.Cl.Cl
C10H18Cl2N2
MFCD00012482
n,n,n',n'-tetramethyl-p-phenylenediamine dihydrochloride, wurster's reagent dihydrochloride, n,n,n',n'-tetramethyl-1,4-phenylenediamine dihydrochloride, unii-66w8hka51x, n1,n1,n4,n4-tetramethylbenzene-1,4-diamine dihydrochloride, wurster's blue dihydrochloride, 1,4-benzenediamine, n,n,n',n'-tetramethyl-, dihydrochloride, 1,4-benzenediamine, n1,n1,n4,n4-tetramethyl-, hydrochloride 1:2, 1-n,1-n,4-n,4-n-tetramethylbenzene-1,4-diamine dihydrochloride, wursters reagent
1-N,1-N,4-N,4-N-tetramethylbenzene-1,4-diamine;dihydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 211-274-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only