missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N,N',N'-Tetramethyldiaminomethane, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 138400250

 View more versions of this product

Catalog No. AC138400250

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N,N,N',N'-Tetramethyldiaminomethane
51-80-9
100.0
0.7400g/mL
−12°C
98.5% min. (GC)
C5H14N2
(CH3)2NCH2N(CH3)2
25 mL
03,21; 07,359
n,n,n',n'-tetramethyldiaminomethane, bis dimethylamino methane, methanediamine, n,n,n',n'-tetramethyl, tetramethylmethylenediamine, methylenebis dimethylamine, n,n,n',n'-tetramethylmethylenediamine, unii-z870i525ks, methylenediamine, n,n,n',n'-tetramethyl, dimethylamino methyl dimethylamine, dimethyl dimethylamino methyl amine
CN(C)CN(C)C
102.18
102.18
Liquid
99%
98.5
Colorless
85.0°C
Authentic
Glass bottle
1.3995 to 1.4015
MFCD00008328
04, 54
0.74
VGIVLIHKENZQHQ-UHFFFAOYSA-N
N,N,N',N'-tetramethylmethanediamine
5829
99%

Chemical Identifiers

51-80-9
102.18
VGIVLIHKENZQHQ-UHFFFAOYSA-N
5829
CN(C)CN(C)C
C5H14N2
MFCD00008328
n,n,n',n'-tetramethyldiaminomethane, bis dimethylamino methane, methanediamine, n,n,n',n'-tetramethyl, tetramethylmethylenediamine, methylenebis dimethylamine, n,n,n',n'-tetramethylmethylenediamine, unii-z870i525ks, methylenediamine, n,n,n',n'-tetramethyl, dimethylamino methyl dimethylamine, dimethyl dimethylamino methyl amine
N,N,N',N'-tetramethylmethanediamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 200-124-7

RTECSNumber : PA6700000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only