missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N,N',N'-Tetramethylethylenediamine, 99%, extra pure, Thermo Scientific Chemicals

$59.12 - $209.60

Chemical Identifiers

CAS 110-18-9
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.21
InChI Key KWYHDKDOAIKMQN-UHFFFAOYSA-N
Synonym temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
PubChem CID 8037
ChEBI CHEBI:32850
IUPAC Name N,N,N',N'-tetramethylethane-1,2-diamine
SMILES CN(C)CCN(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC138450500
SDS View Documents
Thermo Scientific Chemicals
138450500
50 mL Glass bottle N/A
 
AC138455000
SDS View Documents
Thermo Scientific Chemicals
138455000
500 mL Glass bottle N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

110-18-9
116.21
temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
CHEBI:32850
CN(C)CCN(C)C
C6H16N2
KWYHDKDOAIKMQN-UHFFFAOYSA-N
8037
N,N,N',N'-tetramethylethane-1,2-diamine

Specifications

110-18-9
0.7700g/mL
17°C
98.5% min. (GC)
C6H16N2
(CH3)2NCH2CH2N(CH3)2
04,25
0.77
temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
KWYHDKDOAIKMQN-UHFFFAOYSA-N
N,N,N',N'-tetramethylethane-1,2-diamine
8037
116.21
Extra Pure
N,N,N',N'-Tetramethylethylenediamine
-55.0°C
120.0°C to 122.0°C
Authentic
Glass bottle
1.4169 to 1.4189
50 mL
02,403; 03,284; 04,485; 05,652; 06,576; 07,358; 11,340; 12,477; 14,68; 15,63; 16,55
15,9277
Solubility in water: miscible. Other solubilities: most organic solvents
CN(C)CCN(C)C
116.21
CHEBI:32850
99%
Liquid
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 203-744-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only