Learn More
N,N,N',N'-Tetramethylethylenediamine, 99%
CAS: 110-18-9 | C6H16N2 | 116.208 g/mol
$75.74 - $741.42
Chemical Identifiers
| CAS | 110-18-9 |
|---|---|
| Molecular Formula | C6H16N2 |
| Molecular Weight (g/mol) | 116.208 |
| MDL Number | MFCD00008335 |
| InChI Key | KWYHDKDOAIKMQN-UHFFFAOYSA-N |
| Synonym | temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl |
| PubChem CID | 8037 |
| ChEBI | CHEBI:32850 |
| IUPAC Name | N,N,N',N'-tetramethylethane-1,2-diamine |
| SMILES | CN(C)CCN(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA12536AE
|
Thermo Scientific Chemicals
A12536AE |
100 mL |
Each for $75.74
|
|
|||||
|
AAA12536AP
|
Thermo Scientific Chemicals
A12536AP |
500 mL |
Each for $194.18
|
|
|||||
|
AAA125360F
|
Thermo Scientific Chemicals
A125360F |
2500 mL |
Each for $741.42
|
|
|||||
Description
N,N,N',N'-Tetramethylethylenediamine is used with ammonium persulfate to catalyze the polymerization of acrylamide and bis-acrylamide to get polyacrylamide gels. It is also used for the separation of proteins or nucleic acids. Further, it is employed as a ligand for metal ions like zinc and copper. It is actively involved in the formation of anionic organometallic complex.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N,N′,N′-Tetramethylethylenediamine is used with ammonium persulfate to catalyze the polymerization of acrylamide and bis-acrylamide to get polyacrylamide gels. It is also used for the separation of proteins or nucleic acids. Further, it is employed as a ligand for metal ions like zinc and copper. It is actively involved in the formation of anionic organometallic complex.
Solubility
Miscible with water.
Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, carbon dioxide and copper.
Chemical Identifiers
| 110-18-9 | |
| 116.208 | |
| KWYHDKDOAIKMQN-UHFFFAOYSA-N | |
| 8037 | |
| N,N,N',N'-tetramethylethane-1,2-diamine |
| C6H16N2 | |
| MFCD00008335 | |
| temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl | |
| CHEBI:32850 | |
| CN(C)CCN(C)C |
Specifications
| 110-18-9 | |
| 0.775 | |
| 18°C (62°F) | |
| C6H16N2 | |
| (CH3)2NCH2CH2N(CH3)2 | |
| 100 mL | |
| 1732991 | |
| temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl | |
| KWYHDKDOAIKMQN-UHFFFAOYSA-N | |
| N,N,N',N'-tetramethylethane-1,2-diamine | |
| 8037 | |
| 116.21 | |
| Liquid |
| °C to 55°C | |
| 120°C to 122°C | |
| 99% | |
| 1.418 | |
| MFCD00008335 | |
| UN2372 | |
| Air sensitive; Hygroscopic | |
| Miscible with water. | |
| CN(C)CCN(C)C | |
| 116.208 | |
| CHEBI:32850 | |
| 99% | |
| N,N,N',N'-Tetramethylethylenediamine |
Safety and Handling
GHS H Statement
H225-H314-H302-H332
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302+H332-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1,2-DI-(DIMETHYLAMINO)ETHANE
EINECSNumber : 203-744-6
RTECSNumber : KV7175000
TSCA : Yes
storageNote1 : Hygroscopic
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only